Skip to Content
Merck
All Photos(6)

Key Documents

252549

Sigma-Aldrich

Formaldehyde solution

ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)

Synonym(s):

Formalin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCHO
CAS Number:
Molecular Weight:
30.03
Beilstein:
1209228
MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

vapor density

1.03 (vs air)

vapor pressure

52 mmHg ( 37 °C)
52 mmHg ( 37 °C)

form

liquid

autoignition temp.

572 °F

contains

10-15% Methanol as stabilizer (to prevent polymerization)

concentration

36.5-38.0%
37 wt. % in H2O

impurities

≤0.006 meq/g Titr. acid

ign. residue

≤0.005%

color

APHA: ≤10

refractive index

n20/D 1.3765

solubility

water: soluble

density

1.09 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤5 ppm
sulfate (SO42-): ≤0.002%

cation traces

Fe: ≤5 ppm
heavy metals: ≤5 ppm (by ICP-OES)

suitability

suitable for manufacturing use (Laboratory chemicals)

SMILES string

[H]C([H])=O

InChI

1S/CH2O/c1-2/h1H2

InChI key

WSFSSNUMVMOOMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Formaldehyde (formalin) is produced by oxidation of methanol. It is made of 37% formaldehyde and impurities such as methanol, small amounts of formic acid, aldehydes and ketones. It is used as a denaturant in formaldehyde-agarose gel electrophoresis of RNA.

Application

Formaldehyde solution has been used as a fixing agent to fix cells during immunofluorescence imaging and for cross-linking cells during chromatin immunoprecipitation (ChIP) assay.
Formaldehyde solution has been used for cross-linking/fixing of cells in ChIP (chromatin immunoprecipitation) assay. It has been used for fixing of cells for imaging.

Physical form

This product is a solution of approximately 37% by weight of formaldehyde gas in water.

also commonly purchased with this product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Flam. Liq. 3 - Muta. 2 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 1 - STOT SE 3

Target Organs

Eyes,Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

132.8 °F - closed cup

Flash Point(C)

56 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intracellular protein and nucleic acid measured in eight cell types using deep-ultraviolet mass mapping.
Cheung MC et al.
Cytometry, 83, 540-540 (2013)
Mitochondrial translocation of alpha-synuclein is promoted by intracellular acidification.
Cole NB et al.
Experimental Cell Research, 314, 2076-2076 (2008)
Slug contributes to cancer progression by direct regulation of ERa signaling pathway.
Li Y et al.
International Journal of Oncology, 46, 1461-1461 (2015)
SIRT6 protects against endothelial dysfunction and atherosclerosis in mice.
Xu S et al.
Aging (Albany. NY.), 8, 1064-1064 (2016)
Farrell RE
RNA Methodologies: A Laboratory Guide for Isolation and Characterization (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service