Skip to Content
Merck
All Photos(1)

Key Documents

M2284

Sigma-Aldrich

Manganese(III) acetylacetonate

technical grade

Synonym(s):

2,4-Pentanedione manganese(III) derivative, Manganic acetylacetonate, Mn(acac)3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mn(C5H7O2)3
CAS Number:
Molecular Weight:
352.26
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

reaction suitability

core: manganese
reagent type: catalyst

mp

159-161 °C (dec.) (lit.)

SMILES string

CC(=O)\C=C(\C)O[Mn](O\C(C)=C/C(C)=O)O\C(C)=C/C(C)=O

InChI

1S/3C5H8O2.Mn/c3*1-4(6)3-5(2)7;/h3*3,6H,1-2H3;/q;;;+3/p-3/b3*4-3-;

InChI key

HYZQBNDRDQEWAN-LNTINUHCSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Oxidant used to oxidize phenols, β-dicarbonyl compounds, and thiols.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nishino, H. Tategami, S. et al.
Bulletin of the Chemical Society of Japan, 64, 1800-1800 (1991)
Snider, B. B.; Patricia, J. J. et al.
The Journal of Organic Chemistry, 53, 2137-2137 (1988)
Zhan'ao Tan et al.
Physical chemistry chemical physics : PCCP, 14(42), 14589-14595 (2012-09-28)
A solution-processed vanadium oxide (s-VO(x)) anode buffer layer on an indium-tin-oxide (ITO) electrode was used instead of PEDOT:PSS for improving the stability and photovoltaic performance of the polymer solar cells (PSCs). The s-VO(x) layer was prepared by spin-coating a vanadyl
Nan Tian et al.
Dalton transactions (Cambridge, England : 2003), 39(37), 8613-8615 (2010-08-19)
The design, synthesis, photophysical and significantly improved electrooptical properties of a series of red emitting cyclometalated iridium(iii) complexes containing carbazolyl-acetylacetonate ligands are described.
Zofia Kowalewska et al.
Talanta, 103, 66-74 (2012-12-04)
The aim of this work was to investigate the influence of magnesium acetylacetonate (MgA) on the signal of organic forms of vanadium in xylene solution by graphite furnace atomic absorption spectrometry. MgA alone or mixed with palladium acetylacetonate (PdA) was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service