Skip to Content
Merck
All Photos(2)

Key Documents

517003

Sigma-Aldrich

Iron(III) acetylacetonate

≥99.9% trace metals basis

Synonym(s):

2,4-Pentanedione iron(III) derivative, Fe(acac)3, Ferric acetylacetonate, Iron(III) 2,4-pentanedionate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe(C5H7O2)3
CAS Number:
Molecular Weight:
353.17
Beilstein:
4157960
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.9% trace metals basis

form

powder

reaction suitability

core: iron
reagent type: catalyst

mp

180-182 °C (dec.) (lit.)

density

5.24 g/mL at 25 °C (lit.)

SMILES string

CC(=O)\C=C(\C)O[Fe](O\C(C)=C/C(C)=O)O\C(C)=C/C(C)=O

InChI

1S/3C5H8O2.Fe/c3*1-4(6)3-5(2)7;/h3*3,6H,1-2H3;/q;;;+3/p-3/b3*4-3-;

InChI key

AQBLLJNPHDIAPN-LNTINUHCSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Iron(III) acetylacetonate, also known as tris(acetylacetonato)iron(III) or Fe(acac)3, is an coordination compound that is used as a precursor for the synthesis of iron oxide nanoparticles and thin films, which are important in magnetic materials, catalysis, and electronic applications. Additionally, it is also used as a catalyst in various organic reactions, including polymerization processes and cross-coupling reactions.

Application

Iron(III) acetylacetonate can be used as:     
  • A iron precursor for the synthesis of Fe3O4/carbon composite fibers via forcespinning technique. This composite material used in the formation of high-performance anode materials for lithium-ion batteries.      
  • A precursor for the synthesis of iron-containing metal-organic frameworks (MOFs) for the applications in rechargeable alkali-ion batteries.   
  • An additive to enhance the efficiency of the N-hydroxyphthalimide (NHPI) catalyst in the oxidation of cumene.     
  • A solvent activation agent in the fabrication of polyamide membranes, which are used in reverse osmosis (RO) applications.     
  • As a MOCVD precursor for highly crystalline (Zn,Fe)Fe2O4 films and magnetic property measurements of these films. Iron (III) acetylacetonate may be used as a precursor for the synthesis of water-soluble magnetite nanoparticles, which may find applications in magnetic hyperthermia treatment.      
  • As a MOCVD precursor for highly crystalline (Zn,Fe)Fe2O4 films and magnetic property measurements of these films.

MOCVD precursor for highly crystalline (Zn,Fe)Fe2O4 films and magnetic property measurements of these films.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polymorphous transformations of nanometric iron (III) oxide: a review.
Zboril R, et al.
Chemistry of Materials, 23.14, 3255-3272 (2011)
Studies of magnetite nanoparticles synthesized by thermal decomposition of iron (III) acetylacetonate in tri(ethylene glycol)
Maity D, et al.
Journal of magnetism and magnetic materials, 321(19), 3093-3098 (2009)
Selective Detection of Iron (III) by Rhodamine-Modified Fe3O4 Nanoparticles.
Baodui, et al.
Angewandte Chemie (International Edition in English), 49.27, 4576-4579 (2010)
Arreerat Jiamprasertboon et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(48), 11337-11345 (2019-06-27)
Type I heterojunction films of α-Fe2 O3 /ZnO are reported here as a non-titania based photocatalyst, which shows remarkable enhancement in the photocatalytic properties towards stearic acid degradation under UVA-light exposure (λ=365 nm), with a quantum efficiency of ξ=4.42±1.54×10-4 molecules degraded/photon, which
Hyun-Uk Park et al.
Ultrasonics sonochemistry, 58, 104673-104673 (2019-09-27)
In this work, we introduce composition-tunable core-shell-like PdM@Pt (M = Mn and Fe) nanoparticles (NPs) on carbon support (PdM@Pt/C) synthesized by one-pot sonochemical reactions using high-intensity ultrasonic probe (150 W, 20 kHz, with 13 mm solid probe) and investigate their electrocatalytic performance for oxygen reduction

Articles

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service