Skip to Content
Merck
All Photos(1)

Key Documents

MSMLS

Sigma-Aldrich

Mass Spectrometry Metabolite Library

Supplied by IROA Technologies

Synonym(s):

MSMLS, Mass Spectrometry Metabolite Libarary of Standards, Metabolic Library, Metabolite Library

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.25

packaging

pkg of 7 plates

manufacturer/tradename

IROA Technologies

plate size

96 wells

suitability

suitable for mass spectrometry

application(s)

lipidomics
metabolomics
microbiology

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

MSMLS compounds are conveniently provided at 5μg per well, enough for multiple injections. These are high purity (>95%) compounds supplied in an economical, ready to use format. Compounds are arranged in 7 polypropylene racks. Metabolite descriptors provided with product include: Name, Parent CID, molecular formula, molecular weight, CAS, ChEBI, HMDB ID, PubChem Compound and Substance ID.

MSMLS (Mass Spectrometry Metabolite Library) is a collection of high quality small biochemical molecules used for mass spectrometry metabolomics applications and provides a broad representation of primary metabolism.

Application

Mass Spectrometry Metabolite Library is most commonly used to provide retention times and spectra for key metabolic compounds, help optimize analytical mass spectrometry protocols and qualify and quantify mass spectrometry sensitivity and limit of detection.

Features and Benefits

Over 600 unique small molecule metabolites organized in a 96-well format. To view a complete list of compounds in the library for recent lots, download the plate maps below:

  • MSMLS-LOT-230-23
  • MSMLS-LOT-230-16
  • MSMLS-LOT-230-13

    The complete plate map with Name, parent CID, molecular formula, molecular weight, CAS, ChEBI, HMDB ID, PubChem Compound and Substance ID are provided with the software download.

    Broad metabolite spectrum, key primary metabolites and intermediates covering key metabolic pathways, including the following classes of compounds:
  • Carboxylic acids, amino acids
  • Biogenic amines, polyamines
  • Nucleotides, coenzymes and vitamins
  • Mono- and disaccharides
  • Fatty acids, lipids, steroids and hormones

Suitability

Suitable for manual and automated work flows.

Preparation Note

MSMLS compounds are conveniently provided at 5μg per well, enough for multiple injections. Compounds can either be used as standards and injected individually or a strategy may be employed to pool compounds by row to allow multiple mixtures to be analyzed per injection. For more information on MSMLS, solubilization and preparation notes please refer to the product information sheet at the top of the page.

Other Notes

For additional information on our range of Biochemicals, please complete this form.
To download the software package, click here - MLSDiscovery software.




To view the MLSDiscovery Video visit: MLSDiscovery Tutorial Video.

Recommended products

A new metabolite library kit that will feature over 500 unique small molecule metabolites, containing 1 mg of each metabolite. This library of standards is most commonly used for metabolite qualification and quantification for MS and NMR, functional cellular assays and phenotypic screening. Click here to get more information.

Legal Information

MSMLS is a trademark of IROA Technologies LLC

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Lact. - Muta. 2 - Repr. 1A - Resp. Sens. 1 - Skin Corr. 1 - Skin Sens. 1 - STOT RE 1 - STOT RE 2 - STOT RE 2 Oral - STOT SE 2 - STOT SE 3

Target Organs

Blood, Immune system, Respiratory system, Thyroid,Cardio-vascular system,Kidney

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

Flash Point(F)

-40.0 °F

Flash Point(C)

-40 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julian Pezzatti et al.
Journal of chromatography. A, 1592, 47-54 (2019-01-28)
Since the ultimate goal of untargeted metabolomics is the analysis of the broadest possible range of metabolites, some new metrics have to be used by researchers to evaluate and select different analytical strategies when multi-platform analyses are considered. In this
Fernando Vargas et al.
Rapid communications in mass spectrometry : RCM, 34(10), e8725-e8725 (2020-01-14)
A major hurdle in identifying chemicals in mass spectrometry experiments is the availability of tandem mass spectrometry (MS/MS) reference spectra in public databases. Currently, scientists purchase databases or use public databases such as Global Natural Products Social Molecular Networking (GNPS).
Tobias Depke et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1071, 19-28 (2017-06-24)
Pseudomonas aeruginosa is an important opportunistic pathogen that produces a large arsenal of small molecule virulence factors and quorum sensing signal molecules. The annotation of these secondary metabolites in untargeted, mass spectrometry-based metabolomics is difficult, as many of them cannot
Víctor González-Ruiz et al.
Journal of chromatography. A, 1527, 53-60 (2017-11-07)
This work introduces a strategy for decomposing variable contributions within the data obtained from structured metabolomic studies. This approach was applied in the context of an in vitro human neural model to investigate biochemical changes related to neuroinflammation. Neural cells
A three-minute method for high-throughput quantitative metabolomics and quantitative tracing experiments of central carbon and nitrogen pathways.
Nemkov T, et al.
Rapid Communications in Mass Spectrometry, 31(8), 663-673 (2017)

Protocols

MSMLS™ offers high-purity small biochemical molecules for broad primary metabolism studies in an economical, ready-to-use format.

MSMLS™ offers high-purity small biochemical molecules for broad primary metabolism studies in an economical, ready-to-use format.

MSMLS™ offers high-purity small biochemical molecules for broad primary metabolism studies in an economical, ready-to-use format.

MSMLS™ offers high-purity small biochemical molecules for broad primary metabolism studies in an economical, ready-to-use format.

Related Content

We provide small molecule metabolite libraries and kits with a comprehensive range of primary metabolite compounds covering key metabolic pathways for your metabolomics research.

We provide small molecule metabolite libraries and kits with a comprehensive range of primary metabolite compounds covering key metabolic pathways for your metabolomics research.

We provide small molecule metabolite libraries and kits with a comprehensive range of primary metabolite compounds covering key metabolic pathways for your metabolomics research.

We provide small molecule metabolite libraries and kits with a comprehensive range of primary metabolite compounds covering key metabolic pathways for your metabolomics research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service