Skip to Content
Merck
All Photos(4)

Key Documents

283657

Sigma-Aldrich

Nickel(II) acetylacetonate

95%

Synonym(s):

2,4-Pentanedione nickel(II) derivative, Ni(acac)2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ni(C5H7O2)2
CAS Number:
Molecular Weight:
256.91
Beilstein:
4157970
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

95%

form

solid

reaction suitability

core: nickel
reagent type: catalyst

mp

230 °C (dec.) (lit.)

SMILES string

CC(=O)\C=C(\C)O[Ni]O\C(C)=C/C(C)=O

InChI

1S/2C5H8O2.Ni/c2*1-4(6)3-5(2)7;/h2*3,6H,1-2H3;/q;;+2/p-2/b2*4-3-;

InChI key

BMGNSKKZFQMGDH-FDGPNNRMSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Nickel(II) acetylacetonate, known as bis(acetylacetonato)nickel(II) or Ni(acac)2, is a blue-green crystalline compound soluble in water. It finds application in catalysis, materials synthesis, and organic chemistry, playing a crucial role as a catalyst in various reactions and serving as an important precursor for the development of nickel-based nanomaterials and catalysts used in industrial processes and organic transformations.

Application

Nickel(II) acetylacetonate can be:   
  • Used as a precursor to synthesize Ni-based nanomaterials such as NiO/C nanocomposite and crystalline NiO nanoparticles via different synthetic methods like non-isothermal decomposition and solvothermal method.,·      
  • Used to prepare Ni catalysts such as Nickel(II) complexes, and hierarchical Ni/beta catalysts for various organictransformations.
  • Nickel(II) acetylacetonate has several applications in catalysis: It is used as a catalyst for the polymerization of olefins and transesterification reactions.    
  • Nickel(II) acetylacetonate can be employed as a catalyst to promote Michael additions.

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 1A - Muta. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

428.0 °F

Flash Point(C)

220 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huiming Li et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 24(45), 11748-11754 (2018-06-06)
The phase of nanocrystals has a key role in the modulation of catalytic properties. Uniform and well-crystallized nickel phosphide nanocrystals with controlled phases (Ni5 P4 , Ni2 P, and Ni12 P5 ) and narrow size distributions are synthesized by a
Yueping Zhao et al.
Small (Weinheim an der Bergstrasse, Germany), 15(17), e1900288-e1900288 (2019-03-29)
The catalytic performance of Pt-based catalysts for oxygen reduction reactions (ORR) can generally be enhanced by constructing high-index exposed facets (HIFs). However, the synthesis of Pt alloyed high-index skins on 1D non-Pt surfaces to further improve Pt utilization and stability
Husileng Lee et al.
ChemSusChem, 13(12), 3277-3282 (2020-04-02)
Water oxidation is the bottleneck reaction in artificial photosynthesis. Exploring highly active and stable molecular water oxidation catalysts (WOCs) is still a great challenge. In this study, a water-soluble NiII complex bearing a redox non-innocent tetraamido macrocyclic ligand (TAML) is
Lin Chen et al.
Nature communications, 8, 14136-14136 (2017-01-11)
Incorporating oxophilic metals into noble metal-based catalysts represents an emerging strategy to improve the catalytic performance of electrocatalysts in fuel cells. However, effects of the distance between the noble metal and oxophilic metal active sites on the catalytic performance have
Bowen Yu et al.
Nanotechnology, 25(32), 325602-325602 (2014-07-24)
Ni-assisted thermal chemical vapor deposition (TCVD) is one of the most common techniques for the growth of carbon nanofibres/nanotubes (CNFs/CNTs). However, some fundamental issues related to the catalytic growth of CNFs/CNTs, such as the low-limit growth temperature, the limiting steps

Articles

Magnetic nanoparticles have attracted tremendous attention due to their novel properties and their potential applications in magnetic recording, magnetic energy storage and biomedicine.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service