Skip to Content
Merck
All Photos(1)

Key Documents

705314

Sigma-Aldrich

Poly(glycidyl methacrylate)

Mn 10,000-20,000

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C7H10O3]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

Quality Level

mol wt

Mn 10,000-20,000

refractive index

n20/D 1.381

mp

274-280 °C

SMILES string

O1C(C1)COC(=O)C(=C)C

InChI

1S/C7H10O3/c1-5(2)7(8)10-4-6-3-9-6/h6H,1,3-4H2,2H3

InChI key

VOZRXNHHFUQHIL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ran Tao et al.
Langmuir : the ACS journal of surfaces and colloids, 28(48), 16580-16587 (2012-11-15)
Initiated chemical vapor deposition (iCVD) of poly(glycidyl methacrylate) from supersaturated monomer vapor is reported. Rapid film growth rates, up to 600 nm/min, were observed. Films grown from supersaturated monomer exhibited distinct surface undulations. The temporal evolution of surface features during
Peng Tao et al.
ACS applied materials & interfaces, 3(9), 3638-3645 (2011-08-10)
Visibly highly transparent indium tin oxide (ITO)/epoxy nanocomposites were prepared by dispersing polyglycidyl methacrylate (PGMA) grafted ITO nanoparticles into a commercial epoxy resin. The oleic acid stabilized, highly crystalline, and near monodisperse ITO nanoparticles were synthesized via a nonaqueous synthetic
Peggy Scholtysek et al.
Langmuir : the ACS journal of surfaces and colloids, 28(44), 15651-15662 (2012-10-11)
Semitelechelic poly(glycerol methacrylate)s having a perfluoroalkyl end group (PGMA(n)-F(9)) were synthesized by ATRP. The interactions of these polymers with different degrees of polymerization with chiral or racemic dipalmitoylphosphatidylcholine (l-DPPC, d-DPPC, or rac-DPPC) monolayers at the air/water interface were studied. Langmuir
Ying Shen et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(43), 13755-13761 (2012-09-19)
The development of magnetic nanoparticles with multiple functions has been an ever-growing field because of their diverse applications in drug delivery, biosensing, cell labeling, and so on. In this study, a facile method was developed to construct multifunctional magnetic nanocomposites.
Y Hu et al.
ACS applied materials & interfaces, 5(3), 703-712 (2012-12-29)
The biocleavable star-shaped vectors (CD-SS-PGEAs) consisting of nonionic β-cyclodextrin (β-CD) cores and disulfide-linked low-molecular-weight poly(glycidyl methacrylate) (PGMA) derivative arms with plentiful flanking secondary amine and hydroxyl groups were successfully proposed for highly efficient gene delivery. A simple two-step method was

Articles

The present publication focuses on synthesis and characterization of nanothick, chemically grafted polymer films (polymer brushes) on various inorganic and polymeric substrates.

The present publication focuses on synthesis and characterization of nanothick, chemically grafted polymer films (polymer brushes) on various inorganic and polymeric substrates.

The present publication focuses on synthesis and characterization of nanothick, chemically grafted polymer films (polymer brushes) on various inorganic and polymeric substrates.

The present publication focuses on synthesis and characterization of nanothick, chemically grafted polymer films (polymer brushes) on various inorganic and polymeric substrates.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service