Skip to Content
Merck
All Photos(1)

Documents

779342

Sigma-Aldrich

Glycidyl methacrylate

≥97.0% (GC)

Synonym(s):

GMA, 2,3-Epoxypropyl methacrylate, Methacrylic acid 2,3-epoxypropyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10O3
CAS Number:
Molecular Weight:
142.15
Beilstein:
2506
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (GC)

form

liquid

contains

~0.01% hydrochinone monomethylether as stabilizer

refractive index

n20/D 1.449 (lit.)
n20/D 1.450

bp

189 °C (lit.)

density

1.075 g/mL at 20 °C
1.042 g/mL at 25 °C (lit.)

SMILES string

CC(=C)C(=O)OCC1CO1

InChI

1S/C7H10O3/c1-5(2)7(8)10-4-6-3-9-6/h6H,1,3-4H2,2H3

InChI key

VOZRXNHHFUQHIL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Glycidyl methacrylate (GMA) is an epoxy-functional monomer generally used to derivatize polymers. It can also undergo polymerization to form poly (glycidyl methacrylate).

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Muta. 2 - Repr. 1B - Skin Corr. 1C - Skin Sens. 1 - STOT RE 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of glycidyl methacrylate (GMA) on the thermal, mechanical and morphological property of biodegradable PLA/PBAT blend and its nanocomposites.
Kumar M, et al.
Bioresource Technology, 101(21), 8406-8415 (2010)
Biologically active polymers: synthesis and antimicrobial activity of modified glycidyl methacrylate polymers having a quaternary ammonium and phosphonium groups.
Kenawy E-R, et al.
Journal of Controlled Release : Official Journal of the Controlled Release Society, 50(1-2), 145-152 (1998)
Synthesis, characterization, and polymerization of glycidyl methacrylate derivatized dextran.
van Dijk-Wolthuis WNE, et al.
Macromolecules, 28(18), 6317-6322 (1995)
Congying Gu et al.
Electrophoresis, 32(19), 2727-2737 (2011-10-11)
Glycidyl methacrylate-bonded β-cyclodextrin (GMA-β-CD) is synthesized as a new chiral monomer by direct chemical bonding with GMA using a fast and simple alternative procedure. Next, rigid and homogenous monolithic columns were prepared by polymerization of GMA-β-CD monomer with ethylene dimethacrylate
Laura Sola et al.
Journal of chromatography. A, 1270, 324-329 (2012-12-04)
Capillary electrophoresis (CE) is one of the most powerful techniques for the separation of biomolecules. However, the separation efficiency of proteins in CE is often compromised by their tendency to interact with the silanol groups on the surface of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service