Skip to Content
Merck
All Photos(1)

Key Documents

475696

Sigma-Aldrich

Poly(ethylene glycol) diglycidyl ether

average MN 500, cross-linking reagent amine reactive, glycidyl

Synonym(s):

Polyethylene glycol, Diepoxy PEG, PEG diglycidyl ether, Polyoxyethylene bis(glycidyl ether)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H5O2-(C2H4O)n-C3H5O
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

Product Name

Poly(ethylene glycol) diglycidyl ether, average Mn 500

mol wt

average Mn 500

reaction suitability

reagent type: cross-linking reagent
reactivity: amine reactive

refractive index

n20/D 1.47

Ω-end

epoxy

α-end

epoxy

polymer architecture

shape: linear
functionality: homobifunctional

storage temp.

2-8°C

SMILES string

O1C(C1)COCCOCC2OC2

InChI

1S/C8H14O4/c1(9-3-7-5-11-7)2-10-4-8-6-12-8/h7-8H,1-6H2

InChI key

AOBIOSPNXBMOAT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene glycol) diglycidyl ether (PEGDGE) shows highly solubility in water. Hence, it easily undergoes hydrolysis followed by ring cleavage reaction in aqueous solution, yielding hydroxyl group. PEGDGE combines with proteins covalently or non-covalently. PEGDGE is widely used in chemical industries for cross linking and surface modifier.

Application

The high solubility of PEGDGE has been successfully employed to immobilize glucose oxidase, d-amino acid oxidase and glutamate oxidase. It may be used as a component for the development of microelectrode biosensors to detect hydrogen peroxide and nitric oxide.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

386.6 °F - closed cup

Flash Point(C)

197.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Direct electrochemistry and electrocatalysis of hemoglobin on a glassy carbon electrode modified with poly (ethylene glycol diglycidyl ether) and gold nanoparticles on a quaternized cellulose support. A sensor for hydrogen peroxide and nitric oxide.
Li F,et al.
Microchimica Acta, 1-9 (2014)
High performance organic solvent nanofiltration membranes: Development and thorough testing of thin film composite membranes made of polymers of intrinsic microporosity (PIMs)
Fritsch D, et al.
Journal of Membrane Science, 401, 222-231 (2012)
Dilu G Mathew et al.
Nano letters, 20(2), 820-828 (2019-09-20)
Tumor-derived extracellular vesicles (tdEVs) are attracting much attention due to their essential function in intercellular communication and their potential as cancer biomarkers. Although tdEVs are significantly more abundant in blood than other cancer biomarkers, their concentration compared to other blood
Oylum Colpankan Gunes et al.
Journal of biomaterials applications, 35(4-5), 515-531 (2020-07-01)
The objective of the study was to produce three-dimensional and porous nanofiber reinforced hydrogel scaffolds that can mimic the hydrated composite structure of the cartilage extracellular matrix. In this regard, wet-electrospun poly(3-hydroxybutyrate-co-3-hydroxyvalerate) (PHBV) nanofiber reinforced carboxymethyl chitosan-silk fibroin (PNFs/CMCht-SF) hydrogel

Articles

Scaffold patterning with poly(ethylene glycol)-based hydrogels for cell presence in 2D and 3D environments on photoactive substrates.

Scaffold patterning with poly(ethylene glycol)-based hydrogels for cell presence in 2D and 3D environments on photoactive substrates.

Scaffold patterning with poly(ethylene glycol)-based hydrogels for cell presence in 2D and 3D environments on photoactive substrates.

Scaffold patterning with poly(ethylene glycol)-based hydrogels for cell presence in 2D and 3D environments on photoactive substrates.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service