Skip to Content
Merck
All Photos(2)

Key Documents

472344

Sigma-Aldrich

Poly(4-vinylpyridine)

average Mw ~60,000

Synonym(s):

4-Vinylpyridine homopolymer, 4-Vinylpyridine polymer, Poly(4-pyridylethylene), Poly(p -vinylpyridine)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C7H7N)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

mol wt

average Mw ~60,000

transition temp

Tg 137 °C (onset, annealed)

solubility

DMF, acetic acid and lower alcohols: soluble

SMILES string

n1ccc(cc1)C=C

InChI

1S/C7H7N/c1-2-7-3-5-8-6-4-7/h2-6H,1H2

InChI key

KFDVPJUYSDEJTH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(4-vinylpyridine) (PVP) is a pyridine containing polymer that is used in the surface modification by immobilizing the atoms present on the surface.

Application

PVP can be used in a variety of applications such as:
  • fabrication of antibacterial surfaces
  • development of pH sensitive systems
  • 3D molecular level ordering systems
  • anti-corrosive coatings
  • dye sensitized solar cells (DSSCs)(90}

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tailored 3D mechanical metamaterials made by dip-in direct-laser-writing optical lithography
Buckmann T, et al.
Advanced Materials, 24(20), 2710-2714 (2012)
Poly (4-vinylpyridine) and poly (4-vinylpyridine poly-3-oxide ethylene) as corrosion inhibitors for Cu60-Zn40 in 0.5 M HNO3
Abed Y, et al.
Anti-Corrosion Methods and Materials, 48(5), 304-308 (2001)
Laura Maggini et al.
ChemSusChem, 6(2), 367-373 (2012-12-15)
Herein, we report the first example of a supramolecular carbon nanotube (CNT)-based magnetic depolluting agent for divalent metal ion (M(2+)) removal from aqueous solutions. In particular, magnetic multi-walled carbon nanotubes (m-MWCNTs) coated with poly(vinylpyridine) (PVPy) self-aggregate in aqueous solutions that
Jinxin Cao et al.
Journal of colloid and interface science, 381(1), 137-142 (2012-06-09)
We report here a novel protein sequestration method using polymeric colloidal particles prepared by emulsion polymerization with reactive block copolymers. Specifically, poly(4-vinyl-N-ethylpyridine bromide)-block-polymethylacrylate block copolymers were synthesized from reversible addition-fragmentation chain transfer polymerization and used as emulsifiers for emulsion polymerization
Ultrafiltration membranes composed of highly cross-linked cationic polymer gel: the network structure and superior separation performance.
Qifeng Wang et al.
Advanced materials (Deerfield Beach, Fla.), 23(17), 2004-2008 (2011-03-18)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service