Skip to Content
Merck
All Photos(1)

Key Documents

63409

Sigma-Aldrich

Sodium malonate dibasic

≥97.0% (NT)

Synonym(s):

Disodium malonate, Malonic acid disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2(COONa)2
CAS Number:
Molecular Weight:
148.03
Beilstein:
3917013
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (NT)

form

solid

SMILES string

[Na+].[Na+].[O-]C(=O)CC([O-])=O

InChI

1S/C3H4O4.2Na/c4-2(5)1-3(6)7;;/h1H2,(H,4,5)(H,6,7);;/q;2*+1/p-2

InChI key

PRWXGRGLHYDWPS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Sodium malonate dibasic can be use in the synthesis of sodium carbonate and sodium oxalate via wet oxidation and catalytic wet oxidation. It can also be used to synthesize malonato-bridged manganese(II)complexes like [Mn(mal)(H2O)2]n and [Mn2(mal)2(4,4′-bipy)(H2O)2]n,(bipy =bipyridine, mal =malonate), which have the tendency to show weak anti-ferromagnetic effect.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Magneto- structural correlations in 2D and 3D extended structures of manganese (II)- malonate systems"
Maji KT, et al.
Inorganic Chemistry, 42(03), 709-716 (2003)
"Selective organic removal from the alumina industrial liquor: Wet oxidation and catalytic wet oxidation of disodium malonate"
Eyer S , et al.
Industrial & Engineering Chemistry Research, 41(05), 1166-1170 (2002)
Matthias Bauwens et al.
PloS one, 7(6), e38428-e38428 (2012-06-22)
In vivo pH imaging has been a field of interest for molecular imaging for many years. This is especially important for determining tumor acidity, an important driving force of tumor invasion and metastasis formation, but also in the process of
Anna Shevchenko et al.
International journal of pharmaceutics, 436(1-2), 403-409 (2012-07-04)
Cocrystallization and salt formation have been shown to entail substantial promise in tailoring the physicochemical properties of drug compounds, in particular, their dissolution and hygroscopicity. In this work, we report on the preparation and comparative evaluation of a new cocrystal
Adriana Wilhelm et al.
Journal of medicinal chemistry, 55(22), 9817-9830 (2012-10-31)
The protein kinase C-related kinase 2 (PRK2)-interacting fragment (PIF) pocket of phosphoinositide-dependent kinase-1 (PDK1) was proposed as a novel target site for allosteric modulators. In the present work, we describe the design, synthesis, and structure-activity relationship of a series of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service