Skip to Content
Merck
All Photos(3)

Key Documents

223433

Sigma-Aldrich

Sodium oxalate

ACS reagent, ≥99.5%

Synonym(s):

Ethanedioic acid sodium salt, Oxalic acid disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOOCCOONa
CAS Number:
Molecular Weight:
134.00
Beilstein:
3631622
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NB.24
grade:
ACS reagent
form:
powder

grade

ACS reagent

description

neutrality passes test

Assay

≥99.5%

form

powder

impurities

hot H2SO4, passes test (darkened)
≤0.005% insolubles

loss

≤0.01% loss on drying

anion traces

chloride (Cl-): ≤0.002%
sulfate (SO42-): ≤0.002%

cation traces

Fe: ≤0.001%
K: ≤0.005%
NH4+: ≤0.002%
heavy metals (as Pb): ≤0.002%

SMILES string

[Na+].[Na+].[O-]C(=O)C([O-])=O

InChI

1S/C2H2O4.2Na/c3-1(4)2(5)6;;/h(H,3,4)(H,5,6);;/q;2*+1/p-2

InChI key

ZNCPFRVNHGOPAG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sodium oxalate is a water soluble inorganic salt mainly used for the standardization of permanganate solution during titration.

Application

Sodium oxalate may be used as a dispersant and a coordination agent for the electrooxidation of formic acid using a palladium nanoparticle catalyst dispersed on the purified multi-walled carbon nanotubes (P-MWCNTs). In combination with sodium hydroxide, it may be used to mediate the preparation of Cu/ZrO2 catalyst from copper nitrite and zirconium nitrate solution mixture via coprecipitation method.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sodium Hydroxide-Sodium Oxalate-Assisted Co-Precipitation of Highly Active and Stable Cu/ZrO2 Catalyst in the Partial Oxidation of Methanol to Hydrogen.
Chen H, et al.
Catalysis Letters, 131(3-4), 632-642 (2009)
Electrooxidation of formic acid catalyzed by Pd nanoparticles supported on multi-walled carbon nanotubes with sodium oxalate.
Bai Z, et al.
Ionics, 19(3), 543-548 (2013)
Guillem Revilla-López et al.
Langmuir : the ACS journal of surfaces and colloids, 35(36), 11912-11922 (2019-08-03)
Biominerals formed by DNA and calcium oxalate (CaOx) or hydroxyapatite (HAp), the most important and stable phase of calcium phosphate) have been examined and compared using a synergistic combination of computer simulation and experimental studies. The interest of this comparison
The Merck Index: An Encyclopedia of Chemicals, Drugs, and Biologicals
Royal Society of Chemistry null
Taiying Zhang et al.
Carbohydrate polymers, 92(1), 334-344 (2012-12-12)
Dilute oxalic acid pretreatment was applied to maple wood to improve compatibility with downstream operations, and its performance in pretreatment and subsequent enzymatic hydrolysis was compared to results for hydrothermal and dilute hydrochloric and sulfuric acid pretreatments. The highest total

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service