Skip to Content
Merck
All Photos(2)

Key Documents

68714

Supelco

Malonic acid

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Propanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2(COOH)2
CAS Number:
Molecular Weight:
104.06
Beilstein:
1751370
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage condition

(store under argon)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

132-135 °C (dec.) (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

OC(=O)CC(O)=O

InChI

1S/C3H4O4/c4-2(5)1-3(6)7/h1H2,(H,4,5)(H,6,7)

InChI key

OFOBLEOULBTSOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

314.6 °F - closed cup

Flash Point(C)

157 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E A Haan et al.
European journal of pediatrics, 144(6), 567-570 (1986-04-01)
A second child with a more severe deficiency of malonyl CoA decarboxylase is described. He is mildly mentally retarded and presented with vomiting, a seizure, hypoglycaemia and mild metabolic acidosis during a urinary tract infection. The urine contained increased amounts
Anna Shevchenko et al.
International journal of pharmaceutics, 436(1-2), 403-409 (2012-07-04)
Cocrystallization and salt formation have been shown to entail substantial promise in tailoring the physicochemical properties of drug compounds, in particular, their dissolution and hygroscopicity. In this work, we report on the preparation and comparative evaluation of a new cocrystal
Matthias Bauwens et al.
PloS one, 7(6), e38428-e38428 (2012-06-22)
In vivo pH imaging has been a field of interest for molecular imaging for many years. This is especially important for determining tumor acidity, an important driving force of tumor invasion and metastasis formation, but also in the process of
Christian Bonhomme et al.
Journal of the American Chemical Society, 134(30), 12611-12628 (2012-06-29)
Strontium is an element of fundamental importance in biomedical science. Indeed, it has been demonstrated that Sr(2+) ions can promote bone growth and inhibit bone resorption. Thus, the oral administration of Sr-containing medications has been used clinically to prevent osteoporosis
Waree Limwikrant et al.
International journal of pharmaceutics, 431(1-2), 237-240 (2012-04-28)
A new 2/1 carbamazepine (CBZ)/malonic acid (MA) cocrystal polymorph form C was formed using a vibrational rod mill, whereas the known cocrystal polymorph form A was prepared using a ball mill. IR measurements showed that the interaction between CBZ and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service