Skip to Content
Merck
All Photos(1)

Documents

490709

Sigma-Aldrich

Sodium pyruvate-1-13C

99 atom % 13C

Synonym(s):

Pyruvic acid-1-13C sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO13CO2Na
CAS Number:
Molecular Weight:
111.04
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C

Quality Level

form

solid

technique(s)

mass spectrometry (MS): suitable

mp

>300 °C (lit.)

mass shift

M+1

storage temp.

2-8°C

SMILES string

[Na+].CC(=O)[13C]([O-])=O

InChI

1S/C3H4O3.Na/c1-2(4)3(5)6;/h1H3,(H,5,6);/q;+1/p-1/i3+1;

InChI key

DAEPDZWVDSPTHF-FJUFCODESA-M

Application

Sodium pyruvate-1-13C results in sensitivity gains that allow disease tracking and interrogation of cellular metabolism by magnetic resonance. Intravenous administration of hyperpolarized [13C]pyruvate enables real-time detection of metabolism by magnetic resonance spectroscopic imaging (MRSI). It is used as a precursor in estimating many volatile organic compounds for use in studying terrestrial biosphere.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wissam Iali et al.
Angewandte Chemie (International ed. in English), 58(30), 10271-10275 (2019-05-23)
Hyperpolarisation methods that premagnetise agents such as pyruvate are currently receiving significant attention because they produce sensitivity gains that allow disease tracking and interrogation of cellular metabolism by magnetic resonance. Here, we communicate how signal amplification by reversible exchange (SABRE)
Kolby J Jardine et al.
Environmental science & technology, 44(7), 2454-2460 (2010-03-10)
Pyruvic acid, central to leaf carbon metabolism, is a precursor of many volatile organic compounds (VOCs) that impact air quality and climate. Although the pathways involved in the production of isoprenoids are well-known, those of several oxygenated VOCs remain uncertain.
Maria J Gonzalez-Rellan et al.
Nature communications, 12(1), 5068-5068 (2021-08-22)
p53 regulates several signaling pathways to maintain the metabolic homeostasis of cells and modulates the cellular response to stress. Deficiency or excess of nutrients causes cellular metabolic stress, and we hypothesized that p53 could be linked to glucose maintenance. We
Jun Zhao et al.
Cell metabolism, 31(5), 937-955 (2020-04-24)
Cell proliferation and inflammation are two metabolically demanding biological processes. How these competing processes are selectively executed in the same cell remains unknown. Here, we report that the enzyme carbamoyl-phosphate synthetase, aspartyl transcarbamoylase, and dihydroorotase (CAD) deamidates the RelA subunit

Articles

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service