Merck
O-GlcNAcylated p53 in the liver modulates hepatic glucose production.
Nature communications (2021-08-22)
Maria J Gonzalez-Rellan, Marcos F Fondevila, Uxia Fernandez, Amaia Rodríguez, Marta Varela-Rey, Christelle Veyrat-Durebex, Samuel Seoane, Ganeko Bernardo, Fernando Lopitz-Otsoa, David Fernández-Ramos, Jon Bilbao, Cristina Iglesias, Eva Novoa, Cristina Ameneiro, Ana Senra, Daniel Beiroa, Juan Cuñarro, Maria Dp Chantada-Vazquez, Maria Garcia-Vence, Susana B Bravo, Natalia Da Silva Lima, Begoña Porteiro, Carmen Carneiro, Anxo Vidal, Sulay Tovar, Timo D Müller, Johan Ferno, Diana Guallar, Miguel Fidalgo, Guadalupe Sabio, Stephan Herzig, Won Ho Yang, Jin Won Cho, Maria Luz Martinez-Chantar, Roman Perez-Fernandez, Miguel López, Carlos Dieguez, Jose M Mato, Oscar Millet, Roberto Coppari, Ashwin Woodhoo, Gema Fruhbeck, Ruben Nogueiras
ABSTRACT
p53 regulates several signaling pathways to maintain the metabolic homeostasis of cells and modulates the cellular response to stress. Deficiency or excess of nutrients causes cellular metabolic stress, and we hypothesized that p53 could be linked to glucose maintenance. We show here that upon starvation hepatic p53 is stabilized by O-GlcNAcylation and plays an essential role in the physiological regulation of glucose homeostasis. More specifically, p53 binds to PCK1 promoter and regulates its transcriptional activation, thereby controlling hepatic glucose production. Mice lacking p53 in the liver show a reduced gluconeogenic response during calorie restriction. Glucagon, adrenaline and glucocorticoids augment protein levels of p53, and administration of these hormones to p53 deficient human hepatocytes and to liver-specific p53 deficient mice fails to increase glucose levels. Moreover, insulin decreases p53 levels, and over-expression of p53 impairs insulin sensitivity. Finally, protein levels of p53, as well as genes responsible of O-GlcNAcylation are elevated in the liver of type 2 diabetic patients and positively correlate with glucose and HOMA-IR. Overall these results indicate that the O-GlcNAcylation of p53 plays an unsuspected key role regulating in vivo glucose homeostasis.
Sigma-Aldrich
PUGNAc, ≥95% (HPLC)
Sigma-Aldrich
Sodium pyruvate-1-13C, 99 atom % 13C
Sigma-Aldrich
Bovine Serum Albumin, cold ethanol fraction, pH 5.2, ≥96%
Sigma-Aldrich
L-Glutamine–Penicillin–Streptomycin solution, L-glutamine 200 mM, streptomycin 10 mg/mL, penicillin 10,000 units, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
IGEPAL® CA-630, for molecular biology
Sigma-Aldrich
Magna ChIP® A/G Chromatin Immunoprecipitation Kit, Single day chromatin immunoprecipitation (ChIP) kit containing all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using magnetic A/G beads.
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Glucagon, ≥95% (HPLC), powder, synthetic
Sigma-Aldrich
MEM Non-essential Amino Acid Solution (100×), without L-glutamine, liquid, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Minimum Essential Medium Eagle, With Earle′s salts and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Polyethylenimine, branched, average Mw ~25,000 by LS, average Mn ~10,000 by GPC, branched
Sigma-Aldrich
OSMI-1, ≥98% (HPLC)