Merck
  • Deamidation Shunts RelA from Mediating Inflammation to Aerobic Glycolysis.
Deamidation Shunts RelA from Mediating Inflammation to Aerobic Glycolysis.
Cell metabolism (2020-04-24)
Jun Zhao, Mao Tian, Shu Zhang, Alireza Delfarah, Ruoyun Gao, Youliang Rao, Ali Can Savas, Anjie Lu, Larissa Bubb, Xiao Lei, Rosa Moshirian, Wenjie Zhu, Cheng Peng, Taijiao Jiang, Lin Chen, Nicholas A Graham, Pinghui Feng
ABSTRACT
Cell proliferation and inflammation are two metabolically demanding biological processes. How these competing processes are selectively executed in the same cell remains unknown. Here, we report that the enzyme carbamoyl-phosphate synthetase, aspartyl transcarbamoylase, and dihydroorotase (CAD) deamidates the RelA subunit of NF-κB in cancer cells to promote aerobic glycolysis and fuel cell proliferation in tumorigenesis. This post-translational modification switches RelA function from mediating the expression of NF-κB-responsive genes to that of glycolytic enzymes, thus shunting the cell's inflammatory response to aerobic glycolysis. Further, we profiled diverse human cancer cell lines and found that high CAD expression and a subset of RELA mutations correlated with RelA deamidation. And by use of inhibitors of key glycolytic enzymes, we validated the pivotal role of RelA deamidation in tumorigenesis of cancer cell lines. This work illuminates a mechanism by which protein deamidation selectively specifies gene expression and consequent biological processes.
Sigma-Aldrich
(±)-Mevalonolactone, ~97% (titration)
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), crystalline
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Pyrimidine, ≥98.0%
Sigma-Aldrich
Sodium pyruvate-1-13C, 99 atom % 13C
Sigma-Aldrich
Uridine, powder, BioReagent, suitable for cell culture
Roche
Cell Proliferation Kit II (XTT), liquid, pkg of 1 kit, suitable for cell analysis, suitable for tissue culture
Sigma-Aldrich
6-Diazo-5-oxo-L-norleucine, crystalline
Millipore
MUG EC broth, NutriSelect® Plus, suitable for microbiology