Skip to Content
Merck
All Photos(2)

Documents

481033

Sigma-Aldrich

Titanium(III) oxide

−100 mesh, 99.9% trace metals basis

Synonym(s):

Dititanium trioxide, Titanium sesquioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ti2O3
CAS Number:
Molecular Weight:
143.73
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.9% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: titanium

particle size

−100 mesh

density

4.49 g/mL at 25 °C (lit.)

SMILES string

O=[Ti]O[Ti]=O

InChI

1S/3O.2Ti

InChI key

GQUJEMVIKWQAEH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Used to study the bioacceptability of medical implant surfaces.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Titanium dioxide applications: Semiconducting material characteristics and diverse functionalities.

Titanium dioxide applications: Semiconducting material characteristics and diverse functionalities.

Titanium dioxide applications: Semiconducting material characteristics and diverse functionalities.

Titanium dioxide applications: Semiconducting material characteristics and diverse functionalities.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service