Skip to Content
Merck
All Photos(3)

Key Documents

472018

Sigma-Aldrich

Strontium carbonate

≥99.9% trace metals basis

Synonym(s):

Strontium carbonate (SrCO), Strontium monocarbonate 3, Strontium(II) carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SrCO3
CAS Number:
Molecular Weight:
147.63
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
≥99.9% trace metals basis
form:
powder
Pricing and availability is not currently available.

Quality Level

Assay

≥99.9% trace metals basis

form

powder

impurities

≤1000.0 ppm Trace Metal Analysis

density

3.7 g/mL at 25 °C (lit.)

SMILES string

[Sr++].[O-]C([O-])=O

InChI

1S/CH2O3.Sr/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

LEDMRZGFZIAGGB-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Strontium carbonate is a weak base widely used as a starting material in the manufacture of glass cathode ray tubes, colorants in fireworks, phosphors for electroluminescent devices, superconductors, and various Sr-based compounds. It can also be used as a cocatalyst in photocatalytic composites, to expand their photoresponsive range.[1]

Application

Strontium carbonate can be used as a precursor to:
  • Fabricate ceramic composites for n-type thermoelectrics such as SrTiO3–TiO2.[2]
  • Prepare SrCO3/SrO systems for thermochemical energy storage application.[3]

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabrication and thermoelectric properties of SrTiO3?TiO2 composite ceramics
A.P. Zavjalov, et al.
Ceramics International (2022)
Insights into utilization of strontium carbonate for thermochemical energy storage
Paola Ammendola, et al
Renewable Energy, 157, 769-781 (2020)
Visible light responsive strontium carbonate catalyst derived from solvothermal synthesis
Pornnaphat Wichannananon, et al.
Catalysts, 10, 1069-1069 (2020)
Wen-Yu Qian et al.
International journal of nanomedicine, 7, 5781-5792 (2012-11-28)
Strontium carbonate nanoparticles (SCNs), a novel biodegradable nanosystem for the pH-sensitive release of anticancer drugs, were developed via a facile mixed solvent method aimed at creating smart drug delivery in acidic conditions, particularly in tumor environments. Structural characterization of SCNs
Sunkyung Choi et al.
Environmental science & technology, 40(8), 2608-2614 (2006-05-11)
Radioactive strontium (90Sr) is an important constituent of the complex wastes from past nuclear weapons production and has been stored in underground tanks at U.S. DOE sites (e.g., Hanford, WA). Using bulk and microfocused EXAFS spectroscopy, we examined temporal changes

Articles

The prevailing strategies for heat and electric-power production that rely on fossil and fission fuels are having a negative impact on the environment and on our living conditions.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service