Skip to Content
Merck
All Photos(1)

Documents

454710

Sigma-Aldrich

Poly(chlorotrifluoroethylene)

powder

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CF2CF(Cl)]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

powder

dielectric constant

2.6, 1 kHz (ASTM D 150)

hardness

75-85 (Shore D)

parameter

2E-3 cc/sec flow rate (230°C 100 MPa)

transition temp

Tg (DSC) 103 °C (onset)
Tm (DSC) 210 °C (onset)

density

1 g/mL at 25 °C

SMILES string

F\C(F)=C(/F)Cl

InChI

1S/C2ClF3/c3-1(4)2(5)6

InChI key

UUAGAQFQZIEFAH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Valves, seals, packaging films, electroluminescent display panels, bearings where high wear is a problem.

Features and Benefits

Chemical resistance, near-zero moisture absorption, low creep, high optical transparency, high electrical insulating capacity, non-flammable and low coefficient of thermal expansion.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Z Damyanovich et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 144(1), 1-5 (2000-04-28)
A low-temperature, high-power NMR probe head design is described which eliminates the problem of electric arc discharge commonly experienced during radiofrequency pulse cycling in a helium environment. A polychlorotrifluoroethylene (Kel-F) coil former, fitted with a solenoid coil, is heat-shrunk onto
R J Greene et al.
Journal of analytical toxicology, 16(1), 28-32 (1992-01-01)
3.1 Oil, referred to as polychlorotrifluoroethylene (pCTFE), is a polymeric mixture consisting primarily of trimers and tetramers of chlorotrifluoroethylene (CTFE) end-capped with chlorine. Inhalation studies have associated dose-related body weight loss, increased organ weights, and abnormal hepatic enzyme activities with
Issues relating to Aclar 33C.
R E Kingsley et al.
Journal of electron microscopy technique, 12(3), 305-305 (1989-07-01)
E R Kinkead et al.
Toxicology and industrial health, 6(1), 17-32 (1990-01-01)
Halocarbon 27-S (HC 27-S), a polymer of chlorotrifluoroethylene (CTFE), is used as a lubricating oil for pumps in hyperbaric chambers. Although monomeric CTFE has been shown to produce renal lesions in rats, the toxicity of CTFE polymers have not been
Aclar film in biological electron mircoscopy.
E B Masurovsky et al.
Journal of electron microscopy technique, 12(2), 172-173 (1989-06-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service