Skip to Content
Merck
All Photos(2)

Key Documents

H8773

Sigma-Aldrich

Halocarbon oil 27

Synonym(s):

1,1,2-trifluoro-2-chloroethene, Chlorotrifluoroethylene homopolymer

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.75

form

liquid

SMILES string

FC(=C(F)Cl)F

InChI

1S/C2ClF3/c3-1(4)2(5)6

InChI key

UUAGAQFQZIEFAH-UHFFFAOYSA-N

General description

Halocarbon oil 27 is a polymer of chlorotrifluoroethylene (CTFE). It is used to lubricate pumps in hyperbaric chambers.

Application

Halocarbon oil 27 has been used in the microinjection of zebrafish embryos. It has also been used for injury (axotomy) assays.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aparna Ratheesh et al.
Developmental cell, 45(3), 331-346 (2018-05-09)
Migrating cells penetrate tissue barriers during development, inflammatory responses, and tumor metastasis. We study if migration in vivo in such three-dimensionally confined environments requires changes in the mechanical properties of the surrounding cells using embryonic Drosophila melanogaster hemocytes, also called macrophages
Sudarshan Chari et al.
Development (Cambridge, England), 146(19) (2019-09-13)
The early embryos of many animals, including flies, fish and frogs, have unusually rapid cell cycles and delayed onset of transcription. These divisions are dependent on maternally supplied RNAs and proteins including histones. Previous work suggests that the pool size
Analyses in zebrafish embryos reveal that nanotoxicity profiles are dependent on surface-functionalization controlled penetrance of biological membranes
Paatero I, et al.
Scientific reports, 7(1), 8423-8423 (2017)
Axon death pathways converge on axundead to promote functional and structural axon disassembly
Neukomm LJ, et al.
Neuron, 95(1), 78-91 (2017)
Yang Liu et al.
Current biology : CB, 29(13), 2157-2166 (2019-07-02)
Hox genes pattern the anterior-posterior axis of animals and are posited to drive animal body plan evolution, yet their precise role in evolution has been difficult to determine. Here, we identified evolutionary modifications in the Hox gene Abd-B that dramatically

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service