Skip to Content
Merck
All Photos(1)

Key Documents

F9755

Sigma-Aldrich

Fluorinert FC-40

immiscible fluorocarbon oil

Synonym(s):

immiscible fluorocarbon oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.47

form

liquid

density

1.85 g/mL at 25 °C (lit.)

application(s)

hematology
histology

storage temp.

room temp

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C10F22/c11-1(12,3(15,16)5(19,20)7(23,24)9(27,28)29)2(13,14)4(17,18)6(21,22)8(25,26)10(30,31)32

InChI key

BPHQIXJDBIHMLT-UHFFFAOYSA-N

General description

Fluorinert FC-40 is a clear, colorless, non-conductive, and non-flammable liquid that is part of the Fluorinert family of products manufactured by 3M. It is a perfluorinated fluid that has excellent thermal and chemical stability and has a high electric strength, making it suitable for use as a heat transfer fluid. Fluorinert FC-40 has a low boiling point (about 155 °C)It is an immiscible fluorocarbon oil.

Application

Fluorinert FC-40 has been used to evaluate its compatibility with real time quantitative PCR. It has been used in the cytoskeletal contractility assay to deposit samples in dish.
For possible use as a high density chase solution in density gradient studies.

Packaging

Packaged and distributed by Sigma-Aldrich

Other Notes

A trademark for a liquid mixture of completely fluorinated aliphatic compounds.

Legal Information

Fluorinert is a trademark of 3M Company

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dielectrophoretic characterization of cells in a stationary nanoliter droplet array with generated chemical gradients.
Ben-Arye T et al.
Biomedical Microdevices, 17, 91-91 (2015)
Senne Cornelis et al.
Scientific reports, 9(1), 7341-7341 (2019-05-16)
The demand to perform forensic DNA profiling outside of centralized laboratories and on the crime scene is increasing. Several criminal investigations would benefit tremendously from having DNA based information available in the first hours rather than days or weeks. However
Influence of segmenting fluids on efficiency, crossing point and fluorescence level in real time quantitative PCR.
Walsh EJ et al.
Biomedical Microdevices, 8, 59-59 (2006)
Patrick S Stumpf et al.
Communications biology, 3(1), 736-736 (2020-12-06)
Biomedical research often involves conducting experiments on model organisms in the anticipation that the biology learnt will transfer to humans. Previous comparative studies of mouse and human tissues were limited by the use of bulk-cell material. Here we show that
A quantitative analysis of contractility in active cytoskeletal protein networks.
Bendix PM, et al.
Biophysical Journal, 94, 3126-3126 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service