Skip to Content
Merck
All Photos(2)

Key Documents

H8898

Sigma-Aldrich

Halocarbon oil 700

Synonym(s):

Chlorotrifluoroethene homopolymer

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.75

form

liquid

SMILES string

FC(=C(F)Cl)F

InChI

1S/C2ClF3/c3-1(4)2(5)6

InChI key

UUAGAQFQZIEFAH-UHFFFAOYSA-N

General description

Halocarbon oil 700 is used for mounting worms on agarose pads. It facilitates movement of worms for microscopic view. The halocarbon acts as a source of oxygen and blocks desiccation during optical digital embryo sampling.

Application

Halocarbon oil 700 has been used:
  • to facilitate piercing of vitelline membranes with a glass capillary to make small holes on spider embryos
  • in microinjection procedure to track the vesicular stomatitis virus infection in Caenorhabditis elegans
  • for live imaging of embryo sample using Zeiss LSM710 confocal system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Drosophila melanogaster: Practical Uses in Cell and Molecular Biology, 514-514 (1995)
Lunhao Chen et al.
Frontiers in neuroscience, 12, 324-324 (2018-06-06)
Here we described an experimental protocol for in vivo imaging of macropinocytosis and subsequent intracellular events. By microinjection, we delivered fluorescence dextrans together with or without ATPγS into transparent Drosophila melanogaster embryos. Using a confocal microscope for live imaging, we
C. elegans: A Practical Approach, 81-81 (1999)
Infection of Caenorhabditis elegans with Vesicular Stomatitis Virus via Microinjection
Martin A, et al.
Bio-protocol, 7(22) (2017)
Multi-color FISH facilitates analysis of cell-type diversification and developmental gene regulation in the Parasteatoda spider embryo
Akiyama-Oda Y and Oda H
Development, Growth & Differentiation, 58(2), 215-224 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service