Skip to Content
Merck
All Photos(2)

Key Documents

371173

Sigma-Aldrich

Methyl pyruvate

90%, technical grade

Synonym(s):

Pyruvic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCO2CH3
CAS Number:
Molecular Weight:
102.09
Beilstein:
1361953
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

form

liquid

refractive index

n20/D 1.404 (lit.)

bp

134-137 °C (lit.)

solubility

ethanol: soluble 1.1 g/10 mL, clear, colorless to almost colorless

density

1.13 g/mL at 25 °C (lit.)

functional group

ester
ketone

storage temp.

2-8°C

SMILES string

COC(=O)C(C)=O

InChI

1S/C4H6O3/c1-3(5)4(6)7-2/h1-2H3

InChI key

CWKLZLBVOJRSOM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl pyruvate is a pyruvic ester. Surface conditions during Pt/silica EUROPT-1 modified by cinchonidine catalyzed by enantioselective hydrogenation of methyl pyruvate to R-(+)-methyl lactate were investigated. Enantioselective hydrogenation of methyl pyruvate over finely dispersed polyvinylpyrrolidone-stabilized and the corresponding alumina-supported iridium clusters modified with cinchonidine was studied. The molecular orientation and conformation of methyl pyruvate on Ni(111) surface has been investigated in the temperature range 105-220K. Methyl pyruvate is reported to stimulate insulin secretion in vitro and in vivo.

Application

Methyl pyruvate may be used to investigate the adsorption and reactivity of methyl pyruvate on Pt{111}. It may be used in the preparation of dimethyl 2,3-dimethylenebutanedioate.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

116.6 °F - closed cup

Flash Point(C)

47 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Fundamental aspects of enantioselective heterogeneous catalysis: the surface chemistry of methyl pyruvate on Pt {111}.
The Journal of Physical Chemistry B, 104(41), 9696-9703 (2000)
Enantioselective hydrogenation of methyl pyruvate over polymer-stabilized and supported iridium clusters.
Zuo X, et al.
J. Mol. Catal. A: Chem., 147(1), 63-72 (1999)
Young-Ra Cho et al.
Molecular oncology, 12(7), 1203-1215 (2018-05-24)
Matrine is a natural compound extracted from the herb Sophora flavescens Ait which is widely used in traditional Chinese medicine for treating various diseases. Recently, matrine was reported to have antitumor effects against a variety of cancers without any obvious
Orientation and conformation of methyl pyruvate on Ni (111).
Castonguay M, et al.
Journal of the American Chemical Society, 122(3), 518-524 (2000)
Chirag B Godiya et al.
Journal of environmental management, 231, 1012-1020 (2019-01-04)
A big challenge for the civilization in energy saving/waste management can be "the regeneration of monomers from the waste plastics followed by their re-polymerization" using an ideal recycling method. Herein, we investigate the thermal depolymerization of poly(methyl methacrylate) (PMMA) using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service