Skip to Content
Merck
All Photos(2)

Documents

D138401

Sigma-Aldrich

Dimethyl acetylenedicarboxylate

95%

Synonym(s):

Dimethyl 2-butynedioate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCOC≡CCO2CH3
CAS Number:
Molecular Weight:
142.11
Beilstein:
607063
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.447 (lit.)

bp

95-98 °C/19 mmHg (lit.)

density

1.156 g/mL at 25 °C (lit.)

SMILES string

COC(=O)C#CC(=O)OC

InChI

1S/C6H6O4/c1-9-5(7)3-4-6(8)10-2/h1-2H3

InChI key

VHILMKFSCRWWIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyl acetylenedicarboxylate (DMAD) is an ester utilized as a dienophile and a dipolarophile in cycloaddition reactions.

Application

Versatile dienophile used in Diels-Alder reactions.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steven E Wheeler et al.
Journal of the American Chemical Society, 132(10), 3304-3311 (2010-02-18)
Stereoselective Diels-Alder cycloadditions that probe substituent effects in aryl-aryl sandwich complexes were studied experimentally and theoretically. Computations on model systems demonstrate that the stereoselectivity in these reactions is mediated by differential pi-stacking interactions in competing transition states. This allows relative
Yi Li et al.
Polymers, 13(3) (2021-01-28)
Big spherulite structure and high crystallinity are the two main drawbacks of poly(butylene succinate) (PBS) and hinder its application. In this work, a new type of copolyester poly(butylene succinate-co-butylene acetylenedicarboxylate) (PBSAD) is synthesized. With the incorporation of acetylenedicarboxylate (AD) units
Dimethyl acetylenedicarboxylate: A versatile tool in organic synthesis
CG Neochoritis, et al.
Synthesis, 537-585 (2014)
Qiuping Ding et al.
The Journal of organic chemistry, 74(2), 921-924 (2008-12-06)
Tandem electrophilic cyclization-[3+2] cycloaddition-rearrangement reactions of 2-alkynylbenzaldoximes, DMAD, and bromine are described, which afford the unexpected isoquinoline-based azomethine ylides in good to excellent yields. The products could be further elaborated via palladium-catalyzed cross-coupling reactions to generate highly functionalized isoquinoline-based stable
Shunmin He et al.
Nature communications, 10(1), 2219-2219 (2019-05-19)
A long-standing question in the field of embryogenesis is how the zygotic genome is precisely activated by maternal factors, allowing normal early embryonic development. We have previously shown that N6-methyladenine (6mA) DNA modification is highly dynamic in early Drosophila embryos

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service