Skip to Content
Merck
All Photos(4)

Key Documents

316393

Sigma-Aldrich

Sodium triacetoxyborohydride

greener alternative

97%

Synonym(s):

STAB

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COO)3BHNa
CAS Number:
Molecular Weight:
211.94
Beilstein:
4047608
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

reaction suitability

reagent type: reductant

greener alternative product score

old score: 5
new score: 1
Find out more about DOZN™ Scoring

greener alternative product characteristics

Waste Prevention
Design for Energy Efficiency
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

116-120 °C (dec.) (lit.)

greener alternative category

SMILES string

[Na+].CC(=O)O[BH-](OC(C)=O)OC(C)=O

InChI

1S/C6H10BO6.Na/c1-4(8)11-7(12-5(2)9)13-6(3)10;/h7H,1-3H3;/q-1;+1

InChI key

HHYFEYBWNZJVFQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium triacetoxyborohydride (NaBH(OAc)3) is particularly effective in reductive aminations due to its large scope, mildness, and selectivity. It is preferred to sodium cyanoborohydride(NaBH3CN) in many applications due to reduced toxicity of the side products formed, and better yields and reproducibility during synthesis.
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives. This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Waste Prevention”, “Design for Energy Efficiency” and “Use of Renewable Feedstocks”. Click here to view its DOZN scorecard.

Application

Sodium triacetoxyborohydride or [NaBH(OAc)3] can be used as a reagent:
  • In the reductive amination of ketones and aldehydes.
  • To prepare N-benzyl-γ-valerolactam by reacting with methyl 4-oxopentanoate and benzylamine via reductive amination/lactamization.
  • To reduce imines and enamines to corresponding amines.
  • To reduce quinolines and isoquinolines to corresponding tetrahydro derivatives.
  • In the hydroboration of alkenes.
  • To synthesize nitroxide biradicals for creating high relaxivity terminal groups linkage to dendrimers.
Applications using Sodium triacetoxyborohydride

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Sol. 1 - Repr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Use of Sodium Triacetoxyborohydride in Reductive Amination of Ketones and Aldehydes
Abdel-Magid et al.
ACS Symp. Ser., 641, 201-216 (1996)
A review on the use of sodium triacetoxyborohydride in the reductive amination of ketones and aldehydes
Abdel-Magid AF and Mehrman SJ
Organic Process Research & Development, 10(5), 971-1031 (2006)
Methods of enhancement of reactivity and selectivity of sodium borohydride for applications in organic synthesis
Periasamy M and Thirumalaikumar M
Journal of Organometallic Chemistry, 609(1-2), 137-151 (2000)
Ethan M McBride et al.
Forensic science international, 288, 278-282 (2018-05-23)
Impurity profiling has been used as a useful tool for analyzing nearly every drug class currently known on the illicit market. Impurities present within seized samples have the potential to determine source of origin, route of synthesis used, as well
Reductive Amination of Aldehydes and Ketones with Sodium Triacetoxyborohydride. Studies on Direct and Indirect Reductive Amination Procedures(1)
Abdel-Magid AF, Carson KG, Harris BD, Maryanoff CA, Shah RD.
The Journal of Organic Chemistry, 61(11), 3849-3862 (1996)

Articles

Sodium triacetoxyborohydride

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service