Skip to Content
Merck
All Photos(1)

Key Documents

E43101

Sigma-Aldrich

Ethyl chlorooxoacetate

98%

Synonym(s):

mono-Ethyl oxalyl chloride, Ethyl chloroglyoxylate, Monoethyl oxalyl chloride, Oxalic acid monoethyl ester chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCOCOOCH2CH3
CAS Number:
Molecular Weight:
136.53
Beilstein:
506725
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

reaction suitability

reagent type: oxidant

refractive index

n20/D 1.416 (lit.)

bp

135 °C (lit.)

density

1.222 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C(Cl)=O

InChI

1S/C4H5ClO3/c1-2-8-4(7)3(5)6/h2H2,1H3

InChI key

OWZFULPEVHKEKS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethyl chlorooxoacetate can be used for the synthesis of:
  • α-keto esters.
  • Functionalized 3-pyrolin-2-ones.
  • Substituted arylglyoxylic acids via Friedel–Crafts acylation.
  • Substituted 9,10-phenanthrenequinones.
  • Quinoxalinone derivatives.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

105.8 °F - closed cup

Flash Point(C)

41 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

A one-pot synthesis of functionalised 3-pyrolin-2-ones by a four-component reaction between triphenylphosphine, primary amines, dimethyl acetylenedicarboxylate and ethyl chlorooxoacetate.
Anary-Abbasinejad M, et al.
J. Chem. Res. (M), 2007(10), 574-574 (2007)
A Convenient Synthesis of alpha-Keto Esters Using Ethyl 2-Pyridyl Oxalate.
Lee, Jae In
J. Korean Chem. Soc., 48(1), 103-103 (2004)
Synthesis of arylglyoxylic acids and their collision-induced dissociation.
Wadhwa K, et al.
Synthetic Communications, 38(24), 4434-4444 (2008)
Shuhua Cao et al.
Dalton transactions (Cambridge, England : 2003), 49(34), 11851-11858 (2020-07-24)
A bimetallic Cu(ii) complex as a novel antitumor chemodynamic therapy agent with glutathione (GSH) depletion properties is successfully synthesized and well characterized. In tumor cells, the Cu2+ ions of the complex are reduced to Cu+ ions by GSH and then
Microwave assisted synthesis of novel imidazo [2, 1-b] thiazole derivative attached to quinoxalinones.
Mukherjee C, et al.
Tetrahedron Letters, 53(45), 6008-6014 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service