Skip to Content
Merck
All Photos(3)

Key Documents

323829

Sigma-Aldrich

Methyl Green

zinc chloride salt, <0.5% crystal violet, Dye content 80 %

Synonym(s):

Ethyl Green

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C27H35BrClN3 · ZnCl2
CAS Number:
Molecular Weight:
653.24
Colour Index Number:
42590
Beilstein:
3914106
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

Assay

>75.0% (HPLC)

form

solid

composition

Dye content, 80%

technique(s)

titration: suitable

impurities

<0.5% crystal violet

pH

0.2-1.8, yellow to blue

mp

>300 °C (lit.)

solubility

water: 1 mg/mL

λmax

420 nm (2nd)
635 nm

ε (extinction coefficient)

≥14000 at 311-317 in water at 0.005 g/L
≥50000 at 629-635 in water at 0.005 g/L
≥9000 at 420-426 in water at 0.005 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Cl-].[Br-].Cl[Zn]Cl.CC[N+](C)(C)c1ccc(cc1)\C(c2ccc(cc2)N(C)C)=C3/C=C\C(C=C3)=[N+](\C)C

InChI

1S/C27H35N3.BrH.3ClH.Zn/c1-8-30(6,7)26-19-13-23(14-20-26)27(21-9-15-24(16-10-21)28(2)3)22-11-17-25(18-12-22)29(4)5;;;;;/h9-20H,8H2,1-7H3;4*1H;/q+2;;;;;+2/p-4

InChI key

RRKSQWOQZFIQKW-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Methyl Green is a triphenylmethane dye which is mainly used in combination with pyronin. It is suitable for the detection of DNA and RNA.

Application

Used in the Methyl Green-Pyronin methods to demonstrate DNA and RNA in tissue sections.

Other Notes

Zinc chloride salt

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

International Review of Cytology, 4 (1955)
Paxton M Moon et al.
Journal of molecular medicine (Berlin, Germany), 93(8), 845-856 (2015-07-04)
Osteoarthritis (OA) is a highly prevalent, disabling joint disease with no existing therapies to slow or halt its progression. Cartilage degeneration hallmarks OA pathogenesis, and pannexin 3 (Panx3), a member of a novel family of channel proteins, is upregulated during
Xiangling Feng et al.
Molecular carcinogenesis, 53(11), 858-870 (2013-08-03)
Previous studies have shown that promoter hypermethylation plays a key role in DLC-1 inactivation in nasopharyngeal carcinoma (NPC). However, DLC-1 mutation in NPC has not been reported, and there remain some discrepancies in methods and results between different groups. Here
Chun-Chi Liang et al.
The Journal of clinical investigation, 124(7), 3080-3092 (2014-06-18)
Lack of a preclinical model of primary dystonia that exhibits dystonic-like twisting movements has stymied identification of the cellular and molecular underpinnings of the disease. The classical familial form of primary dystonia is caused by the DYT1 (ΔE) mutation in
Christodoulos P Pipinikas et al.
Thorax, 69(6), 548-557 (2014-02-20)
Squamous cell carcinoma of the lung is a common cancer with 95% mortality at 5 years. These cancers arise from preinvasive lesions, which have a natural history of development progressing through increasing severity of dysplasia to carcinoma in situ (CIS), and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service