Skip to Content
Merck
All Photos(2)

Key Documents

V5265

Sigma-Aldrich

Crystal Violet Solution

1%, aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H30ClN3
CAS Number:
Molecular Weight:
407.98
Beilstein:
3580948
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

aqueous solution

Quality Level

concentration

1%

technique(s)

microbe id | staining: suitable

color

deep violet, purple

Amax

0.36-0.44 at 589-594 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Cl-].CN(C)c1ccc(cc1)\C(c2ccc(cc2)N(C)C)=C3/C=C\C(C=C3)=[N+](/C)C

InChI

1S/C25H30N3.ClH/c1-26(2)22-13-7-19(8-14-22)25(20-9-15-23(16-10-20)27(3)4)21-11-17-24(18-12-21)28(5)6;/h7-18H,1-6H3;1H/q+1;/p-1

InChI key

ZXJXZNDDNMQXFV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Crystal violet, also known as gentian violet or methyl violet, is the N-hexamethylated derivative of pararosaniline. It is a basic, cationic, acidotropic protein dye used in histological and bacteriological staining. It is well-known for its antibacterial and antifungal properties.

It stains the fatty portions of sebaceous sweat a deep purple color. Crystal violet can also be used to enhance bloody fingerprints. This dye is harmful if inhaled, swallowed or absorbed through skin, contact may cause cancer, severe eye irritation in human beings.

Application

  • Crystal violet is commonly used in Gram staining for the classification of bacteria.
  • It has also been used to detect bacterial adherence to biomedical polymers and to stain DNA in mammalian tissues in Giemsa staining.
  • It has been successfully used to develop a counterion-staining method to detect DNA in agarose gel electrophoresis.
  • Its antimicrobial properties have facilitated its use in the treatment of oral candidiasis, skin infections, and methicillin-resistant Staphylococcus aureus.
  • It has been used as a stain in cell proliferation assays, migration assays, and Boyden chamber assay.
Crystal violet solution for use in Brown-Hopps method for Gram-positive and Gram-negative bacterial staining.

Biochem/physiol Actions

The antimicrobial effects of crystal violet can be attributed to free radical generation and formation of unionized bacterial complexes with the dye. It has also been reported to inhibit protein synthesis and bacterial cell wall formation. It is especially effective against Gram-positive bacteria.

Principle

Crystal violet is the primary stain in Gram staining. It infiltrates the peptidoglycan-rich cell wall of Gram-positive bacteria, and forms entrapped complexes with Gram′s iodine that retain the purple color post-decolorization. Thus, Gram-positive bacteria can be differentiated from Gram-negative bacteria whose peptidoglycan layer is considerably thinner.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Counterion-dye staining method for DNA in agarose gels using crystal violet and methyl orange
Yang, et al.
Electrophoresis, 22(5) (2001)
K Engbaek et al.
Journal of clinical pathology, 32(2), 187-190 (1979-02-01)
Five techniques for Gram staining bacteria in paraffin sections were compared on serial sections of pulmonary tissues from eight bacteriological necropsies. Brown and Hopp's method was the most satisfactory for distinguishing Gram-positive and Gram-negative bacteria. However, this method cannot be
Characterization of potent and selective iodonium-class inhibitors of NADPH oxidases
Lu J, et al.
Biochemical Pharmacology, 43, 25-38 (2017)
Histological and histochemical methods: Theory and practice (5th ed.)
JA Kiernan
Histological and Histochemical Methods: Theory and Practice (2015)
Studies on adsorption of crystal violet dye from aqueous solution onto coniferous pinus bark powder (CPBP)
Ahmad R.
Journal of Hazardous Materials, 171, 767-773 (2009)

Related Content

An overview of the science and practice of bacteriology in clinical diagnostics. Learn more about the application of standard and special stains for microscopic analysis.

An overview of the science and practice of bacteriology in clinical diagnostics. Learn more about the application of standard and special stains for microscopic analysis.

An overview of the science and practice of bacteriology in clinical diagnostics. Learn more about the application of standard and special stains for microscopic analysis.

An overview of the science and practice of bacteriology in clinical diagnostics. Learn more about the application of standard and special stains for microscopic analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service