Skip to Content
Merck
All Photos(1)

Key Documents

67060

Sigma-Aldrich

Methyl Green

zinc chloride salt, for microscopy (Bact., Bot., Hist.)

Synonym(s):

Ethyl Green

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C27H35BrClN3 · ZnCl2
CAS Number:
Molecular Weight:
653.24
Colour Index Number:
42590
Beilstein:
3914106
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

for microscopy (Bact., Bot., Hist.)

Quality Level

form

powder

technique(s)

titration: suitable

pH

0.2-1.8, yellow to blue

mp

>300 °C (lit.)

solubility

water: 0.01 g/10 mL, blue to very deep blue

εmax

≥900 at 624-634 nm in water

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Cl-].[Br-].Cl[Zn]Cl.CC[N+](C)(C)c1ccc(cc1)\C(c2ccc(cc2)N(C)C)=C3/C=C\C(C=C3)=[N+](\C)C

InChI

1S/C27H35N3.BrH.3ClH.Zn/c1-8-30(6,7)26-19-13-23(14-20-26)27(21-9-15-24(16-10-21)28(2)3)22-11-17-25(18-12-22)29(4)5;;;;;/h9-20H,8H2,1-7H3;4*1H;/q+2;;;;;+2/p-4

InChI key

RRKSQWOQZFIQKW-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Methyl green has been used in the histological staining of ovaries. It has also been used as a counterstain in various cells.

Biochem/physiol Actions

Marker dye for DNA in acid and neutral media.
Methyl green is mainly used as a DNA stain in histochemistry. At low concentrations it binds selectively to AT-rich regions of native DNA. However, at high concentrations it is present at all available DNA sites.

Analysis Note

Material may be a mixture of monobromo-monochloro and dichloro ZnCl2 salts.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K R Fox et al.
European journal of histochemistry : EJH, 36(3), 263-270 (1992-01-01)
Methyl green has long been used as a DNA stain in histochemistry. The sequence selective binding of the cationic triphenylmethane dyes methyl green, crystal violet and Malachite green to DNA was investigated by DNAase 1 and micrococcal nuclease footprinting. At
Evaluation of apoptosis as a mechanism of follicular cell atresia in the ovaries of cattle (bos indicus) and buffalo (bubalus bubalis) fetuses.
Santos SSD, et al.
Animal Reproduction, 10, 55-61 (2013)
Monocyte chemoattractant protein-1 in human atheromatous plaques.
Nelken NA, et al.
The Journal of Clinical Investigation, 88, 1121-1121 (1991)
Paxton M Moon et al.
Journal of molecular medicine (Berlin, Germany), 93(8), 845-856 (2015-07-04)
Osteoarthritis (OA) is a highly prevalent, disabling joint disease with no existing therapies to slow or halt its progression. Cartilage degeneration hallmarks OA pathogenesis, and pannexin 3 (Panx3), a member of a novel family of channel proteins, is upregulated during
Chun-Chi Liang et al.
The Journal of clinical investigation, 124(7), 3080-3092 (2014-06-18)
Lack of a preclinical model of primary dystonia that exhibits dystonic-like twisting movements has stymied identification of the cellular and molecular underpinnings of the disease. The classical familial form of primary dystonia is caused by the DYT1 (ΔE) mutation in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service