Skip to Content
Merck
All Photos(3)

Key Documents

284467

Sigma-Aldrich

2-Methoxyethanol

anhydrous, 99.8%

Synonym(s):

Ethylene glycol monomethyl ether, Methyl glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH2CH2OH
CAS Number:
Molecular Weight:
76.09
Beilstein:
1731074
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
bp:
124-125 °C (lit.)
vapor pressure:
6.17 mmHg ( 20 °C)

grade

anhydrous

Quality Level

vapor density

2.62 (vs air)

vapor pressure

6.17 mmHg ( 20 °C)

Assay

99.8%

form

liquid

autoignition temp.

548 °F

expl. lim.

24.5 %

impurities

<0.005% water

evapn. residue

<0.0005%

refractive index

n20/D 1.402 (lit.)

pH

5.0-7.0 (25 °C)

bp

124-125 °C (lit.)

mp

−85 °C (lit.)

density

0.965 g/mL at 25 °C (lit.)

SMILES string

COCCO

InChI

1S/C3H8O2/c1-5-3-2-4/h4H,2-3H2,1H3

InChI key

XNWFRZJHXBZDAG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Methoxyethanol may be used as a solvent to synthesize:
  • [(dF(CF3)ppy)2-Ir-μ-Cl]2 complex, a precursor for synthesizing iridium based photoredox catalysts for use in cross-coupling reactions [dF(CF3)ppy=2-(2,4-difluorophenyl)-5-(trifluoromethyl)pyridine].
  • 3-(2-methoxyethoxy)propene, an intermediate to prepare a water-soluble, non-volatile and odorless tin hydride reagent.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3 - Repr. 1B - STOT RE 2 - STOT SE 1

Target Organs

Immune system, thymus

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Longlong Wang et al.
Nature communications, 11(1), 5889-5889 (2020-11-20)
The space charge layer (SCL) is generally considered one of the origins of the sluggish interfacial lithium-ion transport in all-solid-state lithium-ion batteries (ASSLIBs). However, in-situ visualization of the SCL effect on the interfacial lithium-ion transport in sulfide-based ASSLIBs is still
Water-Soluble Tin Hydride: Tris [3-(2-Methoxyethoxy) Propyl] Stannane
Light J &amp; Breslow RA
Organic Syntheses, 72, 199-199 (1995)
Practical Syntheses of [2,2'-bipyridine]bis[3,5-difluoro-2-[5-(trifluoromethyl)-2-pyridinyl]phenyl]iridium(III)hexafluorophosphate, [Ir{dF(CF3)ppy}2(bpy)]PF6 and [4,4'-bis(tert-butyl)-2,2'-bipyridine]bis[3,5-difluoro-2-[5-(trifluoromethyl)-2-pyridinyl]phenyl]iridium(III)hexafluorophosphate...
Oderinde MS and Johannes JW
Organic Syntheses, 94, 77-77 (2017)
Shauna Robbennolt et al.
Scientific reports, 9(1), 10804-10804 (2019-07-28)
The magnetic properties of mesoporous cobalt ferrite films can be largely tuned by the application of an electric field using a liquid dielectric electrolyte. By applying a negative voltage, the cobalt ferrite becomes reduced, leading to an increase in saturation
Xinjian Shi et al.
Nature communications, 8(1), 701-701 (2017-09-28)
Electrochemical production of hydrogen peroxide (H

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service