Skip to Content
Merck
All Photos(3)

Key Documents

301930

Sigma-Aldrich

2,2-Bis(bromomethyl)-1,3-propanediol

98%

Synonym(s):

1,3-Dibromo-2,2-bis(hydroxymethyl)propane, Dibromoneopentyl glycol, Pentaerythritol dibromide, Pentaerythritol dibromohydrin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2C(CH2Br)2CH2OH
CAS Number:
Molecular Weight:
261.94
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

10 mmHg ( 178 °C)

Quality Level

Assay

98%

form

solid

mp

112-114 °C (lit.)

SMILES string

OCC(CO)(CBr)CBr

InChI

1S/C5H10Br2O2/c6-1-5(2-7,3-8)4-9/h8-9H,1-4H2

InChI key

CHUGKEQJSLOLHL-UHFFFAOYSA-N

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Muta. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K A Treinen et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 13(2), 245-255 (1989-08-01)
The effect of 2,2-bis(bromomethyl)-1,3-propanediol (BMP) on reproduction in Swiss CD-1 mice was evaluated by use of a continuous breeding protocol. BMP was administered in the feed at 0.1, 0.2, and 0.4% concentrations. Both male and female F0 mice (20 pairs
Weixi Kong et al.
Toxicology letters, 222(3), 273-279 (2013-08-21)
2,2-Bis(bromomethyl)-1,3-propanediol (BMP) is a brominated flame retardant used in urethane foams and polyester resins. In a two year dietary study, BMP caused neoplastic lesions at multiple sites including the urinary bladder of both rats and mice. However, liver was not
Shai Ezra et al.
Journal of contaminant hydrology, 86(3-4), 195-214 (2006-05-13)
This study investigates the mechanisms controlling the distribution of 3-bromo-2,2-bis(bromomethyl)propanol (TBNPA) and 2,2-bis(bromomethyl)propan-1,3-diol (DBNPG) in a fractured chalk aquitard. An extensive monitoring program showed a systematic decrease in the TBNPA/DBNPG ratio with distance from the contamination source. Sorption of TBNPA
Shai Ezra et al.
Chemosphere, 79(4), 476-481 (2010-03-02)
The mechanism and kinetics of the spontaneous decomposition of 2,2-bis(bromomethyl)propan-1,3-diol (DBNPG) and its decomposition daughter products were determined in aqueous solution at a temperatures range between 30 and 70 degrees C and pH from 7.0 to 9.5. DBNPG decomposition in
Simone I Hoehle et al.
Drug metabolism and disposition: the biological fate of chemicals, 37(2), 408-416 (2008-11-26)
2,2-Bis(bromomethyl)-1,3-propanediol (BMP) is a brominated flame retardant, previously shown to be a multisite carcinogen in experimental animals. Studies were performed to characterize the dispositional and metabolic fate of BMP after oral or intravenous administration to male Fischer-344 rats. After a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service