Skip to Content
Merck
All Photos(4)

Key Documents

452912

Sigma-Aldrich

Sodium hydride

greener alternative

60 % dispersion in mineral oil

Synonym(s):

Sodium monohydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaH
CAS Number:
Molecular Weight:
24.00
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.22

form

powder (moist, paste-like)

Quality Level

reaction suitability

reagent type: reductant

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

60 % dispersion in mineral oil

mp

800 °C (dec.) (lit.)

greener alternative category

SMILES string

[Na]

InChI

1S/Na.H

InChI key

MPMYQQHEHYDOCL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Direct intercalation into C60 results in the superconducting material (NaH)4C60.
Sodium hydride (60% dispersion in mineral oil) (NaH) can be used for a variety of applications such as:
  • preparation of a plasticizer for the formation of a solid polymer electrolyte (SPE) for optoelectronic application
  • synthesis of heteroaryl thioethers, the common constituents in therapeutic compounds
  • preparation of sodium aluminum hydride (NaAlH4), which can be used as a hydrogen storage material(3)
  • a base and a reducing agent for the deprotonation of alcohols, phenols, amides, ketones, and different functional groups(4)
  • for potential usage in thermal energy storage(5)

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Sol. 1 - Met. Corr. 1 - Skin Corr. 1A - Water-react 1

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elena Burlacu et al.
Nature communications, 8(1), 714-714 (2017-09-30)
While the protein composition of various yeast 60S ribosomal subunit assembly intermediates has been studied in detail, little is known about ribosomal RNA (rRNA) structural rearrangements that take place during early 60S assembly steps. Using a high-throughput RNA structure probing
Enhanced ionic conductivity of semi-IPN solid polymer electrolytes based on star-shaped oligo (ethyleneoxy) cyclotriphosphazenes
He D, et al.
Macromolecules, 45(19), 7931-7938 (2012)
Selective formation of heteroaryl thioethers via a phosphonium ion coupling reaction
Anderson RG, et al.
Tetrahedron, 74(25), 3129-3136 (2018)
S Scintilla et al.
Journal of inorganic biochemistry, 165, 159-169 (2016-11-28)
Since the discovery of cisplatin in the 1960s, other metal complexes have been investigated as potential antitumor agents to overcome the side-effects associated with the administration of the Pt-based drug. In line with our previous research, in this work we
Sodium hydride as a powerful reducing agent for topotactic oxide deintercalation: Synthesis and characterization of the nickel (I) oxide LaNiO2.
Hayward MA, et al.
Journal of the American Chemical Society, 121(38), 8843-8854 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service