Skip to Content
Merck
All Photos(1)

Documents

393657

Sigma-Aldrich

2-Hydroxymethyl-1,3-propanediol

97%

Synonym(s):

Trimethylolmethane, Tris(hydroxymethyl)methane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC(CH2OH)3
CAS Number:
Molecular Weight:
106.12
Beilstein:
1733340
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

mp

63-68 °C (dec.) (lit.)

SMILES string

OCC(CO)CO

InChI

1S/C4H10O3/c5-1-4(2-6)3-7/h4-7H,1-3H2

InChI key

SFRDXVJWXWOTEW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mutlu Sarikaya et al.
Canadian journal of physiology and pharmacology, 98(4), 243-251 (2019-11-20)
Protein kinase C (PKC) and aldose reductase (AR) enzyme activities are increased in diabetes and complications are include retinopathy, nephropathy, and neuropathy. However, the relationship between PKC and AR and the underlying molecular mechanisms is still unclear. We aimed to
Raúl Domínguez-Perles et al.
Food research international (Ottawa, Ont.), 107, 619-628 (2018-03-28)
Phytoprostanes (PhytoPs) and phytofurans (PhytoFs) are prostaglandin-like compounds, contributing to defense signaling and prevention of cellular damage. These plant oxylipins result from autoxidation of α-linolenic acid (ALA) and have been proposed as new bioactive compounds due to their structural analogies
Umar T Twahir et al.
Biochemistry, 55(47), 6505-6516 (2016-11-01)
This contribution describes electron paramagnetic resonance (EPR) experiments on Mn(III) in oxalate decarboxylase of Bacillus subtilis, an interesting enzyme that catalyzes the redox-neutral dissociation of oxalate into formate and carbon dioxide. Chemical redox cycling provides strong evidence that both Mn
Libia Alejandra García-Flores et al.
Redox biology, 11, 586-591 (2017-01-23)
We analyzed biomarkers of lipid peroxidation of the nervous system -F
Iuliana Mihalache et al.
Physical chemistry chemical physics : PCCP, 21(18), 9564-9573 (2019-04-26)
An interconnected graphene network (IGN) structure with excellent photoluminescence (PL) properties was synthesized using a one-pot microwave-assisted hydrothermal carbonization route. The material exhibited intense and excitation-wavelength dependent PL emission located mainly in the UV-blue light range (300-450 nm). The result

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service