Skip to Content
Merck
All Photos(2)

Documents

538256

Sigma-Aldrich

2,2-Dimethyl-1,3-propanediol

99%

Synonym(s):

NPG Glycol, Neopentylglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C(CH2OH)2
CAS Number:
Molecular Weight:
104.15
Beilstein:
605291
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.6 (vs air)

vapor pressure

<0.8 mmHg ( 20 °C)

Assay

99%

autoignition temp.

750 °F

expl. lim.

1.34 %, 149 °F
18.8 %, 177 °F

SMILES string

CC(C)(CO)CO

InChI

1S/C5H12O2/c1-5(2,3-6)4-7/h6-7H,3-4H2,1-2H3

InChI key

SLCVBVWXLSEKPL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,2-Dimethyl-1,3-propanediol readily absorbs moisture on exposure to air.

Application

2,2-Dimethyl-1,3-propanediol may be used in the synthesis of:
  • 2,2-dimethyl-1,3-propanediol cyclic phosphorochloridate
  • 2,2-dimethyl-1,3-propanediol cyclic phenylphosphonate
  • [1′,3′-(2′,2′-dimethylpropylene)]-2-iodo-3-octyl-5-thienylboronate
  • cyclic carbonate
  • 2,2-dimethyl-1,3-propanediol bis(cyclic-2,2-dimethyltrimethylene phosphite)

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kallol Gupta et al.
Nature, 541(7637), 421-424 (2017-01-13)
Oligomerization of membrane proteins in response to lipid binding has a critical role in many cell-signalling pathways but is often difficult to define or predict. Here we report the development of a mass spectrometry platform to determine simultaneously the presence
Synthesis of 6-membered cyclic carbonates from 1, 3-diols and low CO2 pressure: a novel mild strategy to replace phosgene reagents
Gregory GL, et al.
Royal Society of Chemistry Advances, 5.49 , 39404-39408 (2015)
New convenient synthesis of highly regioregular poly (3-octylthiophene) based on the Suzuki coupling reaction
Guillerez S and Bidan G
Synt. Metals, 93.2, 123-126 (1998)
L Sisfontes et al.
Clinica chimica acta; international journal of clinical chemistry, 155(2), 117-122 (1986-03-16)
Patients attending a general hospital for various reasons were screened for raised serum gamma glutamyltransferase (gamma-GT) and positive blood alcohol concentration (BAC). The results served as objective biochemical tests of heavy drinking. Among 419 individuals, 50 (11.9%) met these requirements
Phosphorus-containing derivatives of 2, 2-dimethyl-1, 3-propanediol
McCONNELL RL and Coover Jr HW
The Journal of Organic Chemistry, 24.5, 630-635 (1959)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service