Skip to Content
Merck
All Photos(2)

Documents

213004

Sigma-Aldrich

Benzeneseleninic acid

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SeO2H
CAS Number:
Molecular Weight:
189.07
Beilstein:
1929765
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

121-124 °C (lit.)

SMILES string

O[Se](=O)c1ccccc1

InChI

1S/C6H6O2Se/c7-9(8)6-4-2-1-3-5-6/h1-5H,(H,7,8)

InChI key

WIHKGDVGLJJAMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzeneseleninic acid oxidizes hydrazine or 1,2-disubstituted derivatives to corresponding diazenes.

Application

Benzeneseleninic acid was used as catalyst during the oxidation of sulfides to sulfoxides via ligand coupling on the iodine atom. It was also used in the preparation of 3-methoxy-17a-oxa-D-homoestra-l,3,5(10)-trien-17-one.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

1, 2-Hydrogen shifts in thermal and photic Bamford-Stevens reactions of cyclohexanones. Activation by an endocyclic oxygen.
Olmstead KK and Nickon A.
Tetrahedron, 54(40), 12161-12172 (1998)
Facile oxidation of sulfides to sulfoxides using iodosobenzene and benzeneseleninic acid as a catalyst.
Roh KR, et al.
Tetrahedron Letters, 32(6), 793-796 (1991)
S Zheng et al.
Hua xi yi ke da xue xue bao = Journal of West China University of Medical Sciences = Huaxi yike daxue xuebao, 26(4), 371-374 (1995-12-01)
In view of the negative relationship between selenium and cancer, we designed and synthesized eleven substituted benzeneselenic acids (IVa-k) The antineoplastic activity of the title compounds was evaluated via the tests of inhibition of acetylcholinesterase, HL-60 and K562 in vitro
Oxidation of hydrazines with benzeneseleninic acid and anhydride.
Back TG, et al.
The Journal of Organic Chemistry, 46(8), 1564-1570 (1981)
Craig A Bayse
Journal of inorganic biochemistry, 104(1), 1-8 (2009-10-30)
The toxicity of selenium is a major barrier to its application to the prevention of cancer, cardiovascular disease, and other chronic ailments. Organic seleninic acids, as well as other reducible selenium compounds, have been shown to react with biological sulfhydryls

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service