Skip to Content
Merck
All Photos(1)

Key Documents

384127

Sigma-Aldrich

Diphenyl ditelluride

98%

Synonym(s):

Phenyl ditelluride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2Te2
CAS Number:
Molecular Weight:
409.41
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

mp

65-67 °C (lit.)

SMILES string

[Te]([Te]c1ccccc1)c2ccccc2

InChI

1S/C12H10Te2/c1-3-7-11(8-4-1)13-14-12-9-5-2-6-10-12/h1-10H

InChI key

VRLFOXMNTSYGMX-UHFFFAOYSA-N

General description

Diphenyl ditelluride (PhTe)2 is a versatile reagent used in the organic synthesis. Neurotoxicity by (PhTe)2 depends on the modulation of signaling pathways initiated at the plasma membrane. Comparative toxicological effects of diphenyl diselenide and (PhTe)2 in mice after in vivo administration has been reported. Teratogenic effects of (PhTe)2 on chicken embryo development has been investigated. The oxidative addition of (PhTe)2 to [Pd(PPh3)4] in dichloromethane is reported to yield [Pd6Cl2Te4(TePh)2(PPh3)6]·1/2CH2Cl2.

Application

Diphenyl ditelluride may be used in the assay of organolithium and Grignard reagents.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daiane Francine Meinerz et al.
PeerJ, 2, e290-e290 (2014-04-09)
Organoselenium compounds have been pointed out as therapeutic agents. In contrast, the potential therapeutic aspects of tellurides have not yet been demonstrated. The present study evaluated the comparative toxicological effects of diphenyl diselenide (PhSe)2 and diphenyl ditelluride (PhTe)2 in mice
Carlos André Prauchner et al.
Toxicology mechanisms and methods, 23(9), 660-664 (2013-08-15)
Studies of our group has demonstrated that (PhSe)2 plays some pharmacologic activities. In addition, it is possible that this compound would be an alternative source of organic selenium in animal foods. However, previous works showed that diphenyl diselenide (PhSe)2 and
The oxidative addition of diphenyl diselenide and ditelluride to tetrakis (triphenylphosphine) palladium.
Oilunkaniemi R, et al.
Journal of Organometallic Chemistry, 623(1), 168-175 (2001)
Bruna Comparsi et al.
Toxicology mechanisms and methods, 24(8), 529-535 (2014-05-28)
Diphenyl ditelluride (PhTe)₂ is a versatile molecule used in the organic synthesis and it is a potential prototype for the development of novel biologically active molecules. The mechanism(s) involved in (PhTe)₂ toxicity is(are) elusive, but thiol oxidation of critical proteins
The Journal of Organic Chemistry, 54, 5627-5627 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service