Skip to Content
Merck
All Photos(2)

Documents

541281

Sigma-Aldrich

Methaneseleninic acid

95%

Synonym(s):

Methylseleninic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SeO2H
CAS Number:
Molecular Weight:
127.00
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

mp

128-132 °C (lit.)

SMILES string

C[Se](O)=O

InChI

1S/CH4O2Se/c1-4(2)3/h1H3,(H,2,3)

InChI key

UEQANLFPOFICBH-UHFFFAOYSA-N

General description

Methaneseleninic acid is an oxidizing agent useful in organic synthesis. It is formed as major product during the oxidations of γ-glutamyl-Se-methylselenocysteine and Se-methylselenocysteine.

Application

Methaneseleninic acid may be employed as a ligand in the preparation of lanthanide complexes. It may also be used in the synthesis of a 12-membered macrocycle [(p-MeO-C6H4)2Te(μ-O)-(μ-MeSeO2)(μ4-Cl)]2 by reacting with bis(p-methoxyphenyl)tellurium dichloride.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sreenivasulu Chintala et al.
BMC cancer, 12, 293-293 (2012-07-19)
Clear cell renal cell carcinoma (ccRCC) accounts for more than 80% of the cases of renal cell carcinoma. In ccRCC deactivation of Von-Hippel-Lindau (VHL) gene contributes to the constitutive expression of hypoxia inducible factors 1 and 2 alpha (HIF-α), transcriptional
Maher S Saifo et al.
Molecular cancer, 9, 310-310 (2010-12-04)
Beta-catenin is a multifunctional oncogenic protein that contributes fundamentally to cell development and biology. Elevation in expression and activity of β-catenin has been implicated in many cancers and associated with poor prognosis. Beta-catenin is degraded in the cytoplasm by glycogen
12-Membered Macrocycles Containing Ditelluroxane and Selenotelluroxane Units.
Srungavruksham NK and Baskar V.
European Journal of Inorganic Chemistry, 2012(1), 136-142 (2012)
M Kotrebai et al.
Journal of chromatography. A, 866(1), 51-63 (2000-02-19)
Increasing speciation demands in clinical chemistry, toxicology and nutrition have made the determination of the total elements in a sample inadequate; the amount of an element and the chemical forms in which it is present need to be known. Inductively
Shu Zhang et al.
The Journal of nutritional biochemistry, 22(12), 1137-1142 (2011-02-05)
Selenium, an essential mineral, plays important roles in optimizing human health. Chitosan (CS) is an effective, naturally oriented material for synthesizing nanoparticles with preferable properties such as biocompatibility, biodegradation and resistance to certain enzymes. We have recently shown that cellular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service