Skip to Content
Merck
All Photos(1)

Key Documents

123579

Sigma-Aldrich

Benzenesulfinic acid sodium salt

98%

Synonym(s):

Sodium benzenesulfinate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SO2Na
CAS Number:
Molecular Weight:
164.16
Beilstein:
3598405
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

mp

>300 °C (lit.)

functional group

sulfinic acid

SMILES string

[Na+].[O-]S(=O)c1ccccc1

InChI

1S/C6H6O2S.Na/c7-9(8)6-4-2-1-3-5-6;/h1-5H,(H,7,8);/q;+1/p-1

InChI key

CHLCPTJLUJHDBO-UHFFFAOYSA-M

General description

Benzenesulfinic acid sodium salt when treated with alkynylselenonium salts yields (Z)-β-alkoxyvinylsulfones. It undergoes rhodium-catalyzed desulfinative coupling with aldehydes to yield ketones.

Application

Benzenesulfinic acid sodium salt has been used in synthesis of sulfones by reaction of organic halides and sodium benzenesulfinate using higher alcohols or diols as solvents.It was also used in the synthesis of aryl sulfones via copper-catalyzed cross-coupling with boronic acids and vinyl sulfones via reaction with vicinal dibromides.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis, 1465-1465 (2007)
Sulfones from sodium benzenesulfinate.
Kauffman JM.
Journal of Chemical and Engineering Data, 14(4), 498-499 (1969)
Reactions of alkynylselenonium salts with sodium benzenesulfinate.
Kataoka T, et al.
Tetrahedron Letters, 38(10), 1809-1812 (1997)
Rhodium-Catalyzed Aerobic Coupling between Aldehydes and Arenesulfinic Acid Salts: A Novel Synthesis of Aryl Ketones.
Rao H, et al.
Advanced Synthesis & Catalysis, 353(10), 1701-1706 (2011)
Tetrahedron, 63, 7667-7667 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service