Skip to Content
Merck
All Photos(5)

Key Documents

194131

Sigma-Aldrich

Oxalic acid

98%

Synonym(s):

Ethanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCO2H
CAS Number:
Molecular Weight:
90.03
Beilstein:
385686
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

4.4 (vs air)

Quality Level

vapor pressure

<0.01 mmHg ( 20 °C)

Assay

98%

form

powder or crystals

pH

(1.3 at 9 g/l)

mp

189.5 °C (dec.) (lit.)

solubility

water: soluble 108 g/L at 25 °C

density

1.65 g/cm3 at 18.5 °C

functional group

carboxylic acid

SMILES string

OC(=O)C(O)=O

InChI

1S/C2H2O4/c3-1(4)2(5)6/h(H,3,4)(H,5,6)

InChI key

MUBZPKHOEPUJKR-UHFFFAOYSA-N

Gene Information

human ... SRC(6714)

Looking for similar products? Visit Product Comparison Guide

General description

Oxalic acid is a dicarboxylic acid used in dehydration, elimination, protection, deprotection, condensation, and decarboxylation reactions.[1]

Oxalic acid is present as a potential impurity in some active pharmaceutical ingredients[2].

Application

Oxalic acid was used in determination of oxamic acid, oxalic acid, and oxamide in hydrophobic drug substance matrix by ion-exclusion chromatography[2]. It was used in the synthesis of three-dimensionally ordered macroporous metal oxides or carbonates via templating with polystyrene spheres[3]. It was employed as supporting electrolyte during electrochemical synthesis of polyaniline-polypyrrole composite coatings[4].

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

General synthesis of periodic macroporous solids by templated salt precipitation and chemical conversion.
Yan H, et al.
Chemistry of Materials, 12(4), 1134-1141 (2000)
Characterization of polyaniline-polypyrrole composite coatings on low carbon steel: a XPS and infrared spectroscopy study.
Rajagopalan R and Iroh JO.
Applied Surface Science, 218(1), 58-69 (2003)
Oxalic Acid
Hamilton GS
eEROS (Encyclopedia of Reagents for Organic Synthesis) (2001)
L Yang et al.
Journal of pharmaceutical and biomedical analysis, 22(3), 487-493 (2000-04-15)
Oxalic acid, oxamic acid and oxamide are potential impurities in some active pharmaceutical ingredients (API). The retention and separation of oxalic and oxamic acids are particularly challenging using conventional reversed-phase HPLC due to their high polarity. An ion-exclusion chromatography (IEC)
G M Gadd
Advances in microbial physiology, 41, 47-92 (1999-09-29)
The production of organic acids by fungi has profound implications for metal speciation, physiology and biogeochemical cycles. Biosynthesis of oxalic acid from glucose occurs by hydrolysis of oxaloacetate to oxalate and acetate catalysed by cytosolic oxaloacetase, whereas on citric acid

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service