Skip to Content
Merck
All Photos(1)

Key Documents

41706

Supelco

Oxalic acid

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCO2H
CAS Number:
Molecular Weight:
90.03
Beilstein:
385686
EC Number:
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

4.4 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

Assay

98.0-102.0% (redox titration)

shelf life

limited shelf life, expiry date on the label

mp

189.5 °C (dec.) (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

OC(=O)C(O)=O

InChI

1S/C2H2O4/c3-1(4)2(5)6/h(H,3,4)(H,5,6)

InChI key

MUBZPKHOEPUJKR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Oxalic acid is a low-molecular-weight organic acid that occurs naturally in several plants, fruits, vegetables, food and beverage products.

Application

Oxalic acid may be used as an analytical standard for the determination of the analyte in juice samples, honey, food samples fruits, vegetables, alcoholic and non-alcoholic beverages and cosmetic formulations by various chromatography techniques.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HPLC determination of low molecular weight organic acids in honey with series-coupled ion-exclusion columns
Nozal MJ, et al.
Journal of Liquid Chromatography and Related Technologies, 26(8), 1231-1253 (2003)
HPLC organic acid analysis in different citrus juices under reversed phase conditions
Violeta N, et al.
Notulae Botanicae Horti Agrobotanici Cluj-Napoca , 38(1), 44-48 (2010)
Validation of HPLC stability-indicating method for Vitamin C in semisolid pharmaceutical/cosmetic preparations with glutathione and sodium metabisulfite, as antioxidants
Maia AM, et al.
Talanta, 71(2), 639-643 (2007)
Separation and determination of organic acids and phenolic compounds in fruit juices and drinks by high-performance liquid chromatography
Shui G and Leong LP
Journal of Chromatography A, 977(1), 89-96 (2002)
Simultaneous detection of dehydroascorbic, ascorbic, and some organic acids in fruits and vegetables by HPLC
Nisperos-Carriedo MO, et al.
Journal of Agricultural and Food Chemistry, 40(7), 1127-1130 (1992)

Protocols

Separation of Fumaric acid; Oxalic acid; Citric acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service