Skip to Content
Merck
All Photos(1)

Documents

155756

Sigma-Aldrich

2,2,6,6-Tetramethyl-3,5-heptanedione

98%

Synonym(s):

Dipivaloylmethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCOCH2COC(CH3)3
CAS Number:
Molecular Weight:
184.28
Beilstein:
1447269
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.459 (lit.)

bp

72-73 °C/6 mmHg (lit.)

density

0.883 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)C(=O)CC(=O)C(C)(C)C

InChI

1S/C11H20O2/c1-10(2,3)8(12)7-9(13)11(4,5)6/h7H2,1-6H3

InChI key

YRAJNWYBUCUFBD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,2,6,6-Tetramethyl-3,5-heptanedione is a bidentate ligand used in the synthesis of stable complexes with lanthanide ions.

2,2,6,6-Tetramethyl-3,5-heptanedioneis a stable, anhydrous reagent. It undergoes O-additions and C-additions. In various reactions, it acts as an air-stable ligand for metal catalysts. Furthermore, it serves as a substrate for heterocycles.

Application

2,2,6,6-tetramethyl-3,5-heptanedione was used in the synthesis of α-aryl-β-diketones and dicyanamidobenzene-bridge diruthenium complex.

2,2,6,6-Tetramethyl-3,5-heptanedione used as a ancillary ligand in the synthesis of orange-emitting iridium(III) complex.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

152.6 °F - closed cup

Flash Point(C)

67 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Thermochemistry of 2, 2, 6, 6-tetramethyl-3, 5-heptanedione chelates of lanthanide group elements
Santos Jr, et al.
Polyhedron, 18, 969-977 (1999)
Synthesis and characterization of phenylpyridine derivative containing an imide functional group on an iridium (III) complex for solution-processable orange-phosphorescent organic light-emitting diodes
Hwang J, et al.
Dyes and Pigments, 121, 73-78 (2015)
Solvent extraction of some lanthanides with 2, 2, 6, 6-tetramethyl-3, 5-heptanedione
Sweet TR and Parlett HW
Analytical Chemistry, 40(12), 1885-1888 (1968)
2, 2, 6, 6-Tetramethyl-3, 5-heptanedione
Jurica EA
Encyclopedia of Reagents for Organic Synthesis, Second Edition (2001)
Tetrahedron, 48, 6909-6909 (1992)

Articles

Buchwald phosphine ligands for C-C, C-N, and C-O bond formation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service