T27006
2-Thenoyltrifluoroacetone
99%
Synonym(s):
4,4,4-Trifluoro-1-(2-thienyl)-1,3-butanedione, TTA (extractant), TTB, TTFA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H5F3O2S
CAS Number:
Molecular Weight:
222.18
Beilstein:
168645
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
bp
96-98 °C/8 mmHg (lit.)
mp
40-44 °C (lit.)
storage temp.
2-8°C
SMILES string
FC(F)(F)C(=O)CC(=O)c1cccs1
InChI
1S/C8H5F3O2S/c9-8(10,11)7(13)4-5(12)6-2-1-3-14-6/h1-3H,4H2
InChI key
TXBBUSUXYMIVOS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Reagent for the determination of actinides and lanthanides.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
233.6 °F - closed cup
Flash Point(C)
112 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Vladimir A Kokorekin et al.
Molecules (Basel, Switzerland), 25(18) (2020-09-17)
In this article, we demonstrate how an original effective "metal-free" and "chromatography-free" route for the synthesis of 3-thiocyanatopyrazolo[1,5-a]pyrimidines has been developed. It is based on electrooxidative (anodic) C-H thiocyanation of 5-aminopyrazoles by thiocyanate ion leading to 4-thiocyanato-5-aminopyrazoles (stage 1, yields
Jeffery, G.H. et al.
Vogel's Textbook of Quantitative Chemical Analysis, 170-170 (1989)
Pin-Zhu Qin et al.
Talanta, 80(2), 991-995 (2009-10-20)
A two-probe tandem DNA hybridization assay including capture DNA(1), probe DNA(2), and target DNA(3) was prepared. The long-lived luminescent europium complex doped nanoparticles (NPs) were used as the biomarker. The complex included in the particle was Eu(TTA)(3)(5-NH(2)-phen)-IgG (ETN-IgG), the europium
Hae-Ok Byun et al.
Journal of cellular biochemistry, 104(5), 1747-1759 (2008-04-09)
Mitochondrial complex II defect has recently been implicated in cellular senescence and in the ageing process of which a critical phenotype is retardation and arrest of cellular growth. However, the underlying mechanisms of how complex II defect affects cellular growth
Yajuan Li et al.
Dalton transactions (Cambridge, England : 2003), 39(10), 2554-2562 (2010-02-25)
A series of novel ternary organic-inorganic mesoporous polymeric hybrids TTFA-S16-Eu-PMMA, TTFA-S16-Eu-PMAA, and TTFA-S16-Eu-PVP (TTFA = 2-Thenoyltrifluoroacetone; PMMA = polymethyl methacrylate; PMAA = polymethacrylic acid; PVP = polyvinylpyrrolidone) have been assembled by the Eu(3+) complex covalently attaching to the TTFA directly
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service