Skip to Content
Merck
All Photos(3)

Key Documents

D33454

Sigma-Aldrich

1,3-Diphenyl-1,3-propanedione

98%

Synonym(s):

Dibenzoylmethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CO)2CH2
CAS Number:
Molecular Weight:
224.25
Beilstein:
514910
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

crystals

bp

219-221 °C/18 mmHg (lit.)

mp

77-79 °C (lit.)

SMILES string

O=C(CC(=O)c1ccccc1)c2ccccc2

InChI

1S/C15H12O2/c16-14(12-7-3-1-4-8-12)11-15(17)13-9-5-2-6-10-13/h1-10H,11H2

InChI key

NZZIMKJIVMHWJC-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

1,3-Diphenyl-1,3-propanedione also known as Dibenzoylmethane, a symmetrical ß-diketone with the ability to form strong intra and intermolecular hydrogen bonds is commonly used to produces a Schiff base in reaction with benzidine.

Application


  • A radical-anion chain mechanism following dissociative electron transfer reduction of the model prostaglandin endoperoxide, 1,4-diphenyl-2,3-dioxabicyclo[2.2.1]heptane.: This study explores the unique chemical properties of 1,3-Diphenyl-1,3-propanedione in the context of electron transfer reactions. The research unveils its potential in creating advanced prostaglandin models, crucial for pharmaceutical applications and drug synthesis. This application is especially significant in the context of prostaglandin biosynthesis, offering a pathway to new drug development strategies within the pharmaceutical industry and biochemical research (Magri & Workentin, 2008).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mojtaba Shamsipur et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 77(1), 319-323 (2010-07-17)
A highly sensitive ion-selective bulk optode membrane for sensing UO2(2+) ion based on plasticized poly(vinyl chloride) containing 6,7,9,10,12,13,15,16,23,24,25,26-dodecahydrodibenzo[n,v][1,4,7,10,13,17,20]pentaoxa-diazacyclotricosine-22,27-dione as ionophore, dibenzodylmethane as chromoionophore and sodium tetraphenylborate as an ionic additive was prepared. In addition to its high stability, reproducibility and
Ka Lung Cheung et al.
Carcinogenesis, 31(5), 880-885 (2009-12-05)
Previously, phenethyl isothiocyanate (PEITC) and dibenzoylmethane (DBM) had been shown to inhibit intestinal carcinogenesis in Apc(Min/+) mice. In this study, we investigated the chemopreventive efficacy of PEITC and DBM in the azoxymethane (AOM)-initiated and dextran sodium sulfate (DSS)-promoted colon cancer
Abdou Saad El-Tabl et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 71(1), 90-99 (2008-02-05)
A new series of mono and binuclear Mn(II), Fe(III), Co(II), Ni(II), Cu(II), Zn(II), La(III), Ru(III), Hf(IV), ZrO(II) and UO2(II) complexes of phenylaminodibenzoylhydrazone have been synthesized and characterized by elementals analyses, IR UV-vis spectra, magnetic moments, conductances, thermal analyses (DTA and
Zhaogang Nie et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 72(3), 554-560 (2008-12-17)
Rare-earth complex Sm(DBM)(3)Phen-doped poly(methyl methacrylate) was synthesized and its absorption and luminescence spectra were studied. The relationship between the coordinate environment of rare-earth ions and the Judd-Ofelt intensity parameter Omega(2) was analyzed and the spectroscopic quality factor, Omega(4)/Omega(6), for this
Yuichi Masuda et al.
The journal of physical chemistry. A, 116(18), 4485-4494 (2012-04-19)
Despite the importance of ultrafast (time scale exceeding 10(-11) s) intramolecular proton transfer (PT) events between electronic ground states in solution, experimental determination of the rates of such reactions has not yet been accomplished because of the limitations of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service