Skip to Content
Merck
All Photos(1)

Documents

147281

Sigma-Aldrich

Sodium benzenesulfonate

97%

Synonym(s):

Benzenesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SO3Na
CAS Number:
Molecular Weight:
180.16
Beilstein:
3918459
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39092528
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

solubility

water: soluble

density

1.124 g/mL at 25 °C (lit.)

SMILES string

[Na+].[O-]S(=O)(=O)c1ccccc1

InChI

1S/C6H6O3S.Na/c7-10(8,9)6-4-2-1-3-5-6;/h1-5H,(H,7,8,9);/q;+1/p-1

InChI key

MZSDGDXXBZSFTG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium benzenesulfonate has been used in Raman spectroscopic determination of molecular structural features in sulfonated polystyrene resins. It was used as electrolyte in formation of polypyrrole coatings with varying surface morphology on stainless steel. It was used in the synthesis of 1-butyl-3-propanenitrile imidazolium benzenesulfonate.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of sulfonate-based anions on the physicochemical properties of 1-alkyl-3-propanenitrile imidazolium ionic liquids.
Ziyada AK, et al.
New. J. Chem., 35(5), 1111-1116 (2011)
Raman spectroscopy of sulfonated polystyrene resins.
Edwards HGM, et al.
Vibrational Spectroscopy, 24(2), 213-224 (2000)
Formation of polypyrrole coatings on stainless steel in aqueous benzene sulfonate solution.
Su W and Iroh JO.
Electrochimica Acta, 42(17), 2685-2694 (1997)
Wen Huang et al.
The Journal of organic chemistry, 73(17), 6845-6848 (2008-08-07)
A novel, convenient, and efficient method has been developed for selective synthesis of spirocycle, polycycle, and di- and tetrahydronaphthalene systems from aryl-substituted propargylic alcohols by FeCl3- or TsOH-catalyzed multiple activations of unsaturated C-C bonds and C-H bonds.
Ramasubbu Ramani et al.
Biomacromolecules, 9(5), 1390-1397 (2008-04-19)
We present lamellar self-assembly of cationic poly(L-histidine) (PLH) stoichiometrically complexed with an anionic surfactant, dodecyl benzenesulfonic acid (DBSA), which allows a stabilized conformation reminiscent of polyproline type II (PPII) left-handed helices. Such a conformation has no intrapeptide hydrogen bonds, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service