Skip to Content
Merck
All Photos(1)

Key Documents

PHR1305

Supelco

Sodium dodecylbenzenesulfonate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Dodecylbenzenesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11C6H4SO3Na
CAS Number:
Molecular Weight:
348.48
Beilstein:
4171051
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1623637

API family

sodium dodecylbenzenesulfonate

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

[Na+].CCCCCCCCCCCCc1ccc(cc1)S([O-])(=O)=O

InChI

1S/C18H30O3S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-17-13-15-18(16-14-17)22(19,20)21;/h13-16H,2-12H2,1H3,(H,19,20,21);/q;+1/p-1

InChI key

JHJUUEHSAZXEEO-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium dodecylbenzenesulfonate, commonly known as sodium lauryl phenyl sulfonate, is a linear alkylbenzene sulfonate that belongs to the class of anionic surfactants. Generally, it is used as an emulsifier, detergent, solubilizer and wetting agent in pharmaceutical, personal care, home care and food products.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Sodium dodecylbenzenesulfonate may be used as a pharmaceutical reference standard for the determination of the analyte in raw material, environmental samples, personal care and home care formulations by high performance liquid chromatography.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3264 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simple and rapid determination of linear alkylbenzene sulfonates by in-tube solid-phase microextraction coupled with liquid chromatography
Hirayama Y, et al.
Journal of Health Science, 52(3), 228-236 (2006)
Amended safety assessment of dodecylbenzenesulfonate, decylbenzenesulfonate, and tridecylbenzenesulfonate salts as used in cosmetics
Becker LC, et al.
International Journal of Toxicology, 29, 288S-305S (2010)
Determination of surfactant mixtures in shampoos and detergents by HPLC
Nakamura K and Morikawa Y
Journal of the American Oil Chemists' Society, 61(6), 1130-1135 (1984)
Development of a method by HPLC to determine LAS and its application in anaerobic reactors
Duarte I, et al.
Journal of the Brazilian Chemical Society, 17(7), 1360-1367 (2006)
Specific determination of linear alkylbenzenesulfonates (LAS) in commercial detergents and whole blood by high-performance liquid chromatography with solid-phase extraction
Xue Y, et al.
Journal of Analytical Toxicology, 31(1), 37-43 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service