Merck
  • Controllable one-step synthesis of spirocycles, polycycles, and di- and tetrahydronaphthalenes from aryl-substituted propargylic alcohols.
Controllable one-step synthesis of spirocycles, polycycles, and di- and tetrahydronaphthalenes from aryl-substituted propargylic alcohols.
The Journal of organic chemistry (2008-08-07)
Wen Huang, Pengzhi Zheng, Zhengxing Zhang, Ruiting Liu, Zhenxia Chen, Xigeng Zhou
ABSTRACT
A novel, convenient, and efficient method has been developed for selective synthesis of spirocycle, polycycle, and di- and tetrahydronaphthalene systems from aryl-substituted propargylic alcohols by FeCl3- or TsOH-catalyzed multiple activations of unsaturated C-C bonds and C-H bonds.
Sigma-Aldrich
Benzenesulfonic acid, 98.0% (T)
Sigma-Aldrich
Benzenesulfonic acid, technical grade, 90%
Sigma-Aldrich
Sodium benzenesulfonate, 97%