Skip to Content
Merck
All Photos(3)

Documents

129542

Sigma-Aldrich

Isovaleric acid

99%

Synonym(s):

3-Methylbutanoic acid, 3-Methylbutyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2COOH
CAS Number:
Molecular Weight:
102.13
Beilstein:
1098522
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.38 mmHg ( 20 °C)

Quality Level

Assay

99%

form

liquid

autoignition temp.

824 °F

refractive index

n20/D 1.403 (lit.)

bp

175-177 °C (lit.)

mp

−29 °C (lit.)

density

0.925 g/mL at 20 °C (lit.)

SMILES string

CC(C)CC(O)=O

InChI

1S/C5H10O2/c1-4(2)3-5(6)7/h4H,3H2,1-2H3,(H,6,7)

InChI key

GWYFCOCPABKNJV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isovaleric acid, also known as 3-methylbutanoic acid, is commonly used for ester synthesis and as a building block in organic reactions. It is a volatile organic compound present in vinegar bait and is determined by solid phase microextraction. Isovaleric acid inhibits the synthesis of saturated fatty acids. It is a favorable carbon source for cell growth. Moreover, it is a promising odor indicator.

Application

Isovaleric acid inhibits the synthesis of saturated fatty acids.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

176.0 °F - Pensky-Martens closed cup

Flash Point(C)

80 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaojin Song et al.
Journal of agricultural and food chemistry, 61(41), 9876-9881 (2013-09-24)
Aurantiochytrium is an important docosahexaenoic acid (DHA) producer containing two kinds of fatty acid synthesis pathways, that is, the fatty acid synthase pathway (FAS) for saturated fatty acid synthesis and the polyketide synthase pathway (PKS) for polyunsaturated fatty acid synthesis.
Qinxue Wen et al.
Bioresource technology, 143, 695-698 (2013-07-17)
Microalga Chlorella protothecoides can accumulate high proportion of lipids during the heterotrophic growth with glucose as the carbon source. However, its commercial application is restricted due to the high cost of the carbon source. In this study, the wasted activated
N Qamaruz-Zaman et al.
Waste management (New York, N.Y.), 32(12), 2426-2430 (2012-07-24)
Research was conducted to determine suitable chemical parameters as indicators of odor from decomposing food wastes. Prepared food scraps were stored in 18 l plastic buckets (2 kg wet weight each) at 20 °C and 8 °C to reproduce high
Biological active esters of the isovaleric acid
Suerbaev, et al.
Eurasian Chemico-Technological Journal, 16, 299-302 (2014)
Lingling Wang et al.
Microbiology spectrum, 9(2), e0124621-e0124621 (2021-10-21)
The adenoviral DNA is prevalent in adenotonsillectomy specimens from pediatric patients, though the virus seems to be in latent state. The tonsils are at the forefront of airway entry point and are the first line of defense against airway viral

Articles

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service