Skip to Content
Merck
All Photos(1)

Key Documents

78651

Supelco

Isovaleric acid

analytical standard

Synonym(s):

3-Methylbutanoic acid, 3-Methylbutyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2COOH
CAS Number:
Molecular Weight:
102.13
Beilstein:
1098522
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor pressure

0.38 mmHg ( 20 °C)

Assay

≥98.5% (GC)

autoignition temp.

824 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.3% water

refractive index

n20/D 1.402-1.404
n20/D 1.403 (lit.)

bp

175-177 °C (lit.)

mp

−29 °C (lit.)

density

0.925 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC(C)CC(O)=O

InChI

1S/C5H10O2/c1-4(2)3-5(6)7/h4H,3H2,1-2H3,(H,6,7)

InChI key

GWYFCOCPABKNJV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Isovaleric acid may be used as an analytical reference standard for the quantification of the analyte in the following:
  • Rose apple (Syzygium jambos Alston) fruit using gas chromatography with flame ionization detection (GC-FID), mass spectrometry (GC-MS) and olfactometry/aroma extract dilution analysis (GC-O/AEDA).
  • Sicilian goat cheese using gas chromatography with flame ionization detection (GC-FID) and mass spectrometry (GC-MS).
  • Food products using gas chromatography with flame ionization detection (GC-FID).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

176.0 °F - Pensky-Martens closed cup

Flash Point(C)

80 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative and sensory evaluation of malodour retention of fibre types by use of artificial skin, sweat and radiolabelled isovaleric acid
Hammer RT, et al.
Flavour and Fragrance Journal, 28(4), 238-244 (2013)
Study of the aroma compounds of rose apple (Syzygium jambos Alston) fruit from Brazil
Guedes MC, et al.
European Food Research and Technology, 219(5), 460-464 (2004)
Elena Niccolai et al.
World journal of gastroenterology, 25(36), 5543-5558 (2019-10-03)
An altered (dysbiosis) and unhealthy status of the gut microbiota is usually responsible for a reduction of short chain fatty acids (SCFAs) concentration. SCFAs obtained from the carbohydrate fermentation processes are crucial in maintaining gut homeostasis and their determination in
Giuditta Pagliai et al.
European journal of nutrition, 59(5), 2011-2024 (2019-07-12)
We evaluated the effect of low-calorie mediterranean (MD) and vegetarian (VD) diets on gut microbiome (GM) composition and short-chain-fatty acids (SCFA) production. We performed next generation sequencing (NGS) of 16S rRNA and SCFA analysis on fecal samples of 23 overweight
Determination of acids in Brazilian sugar cane spirits and other alcoholic beverages by HRGC-SPE
Nascimento DFR, et al.
Chromatographia, 48(11-12), 751-757 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service