Skip to Content
Merck
All Photos(3)

Key Documents

153745

Sigma-Aldrich

Hexanoic acid

≥99%

Synonym(s):

Acid C6, Caproic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4COOH
CAS Number:
Molecular Weight:
116.16
Beilstein:
773837
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

4 (vs air)

vapor pressure

0.18 mmHg ( 20 °C)

Assay

≥99%

form

liquid

refractive index

n20/D 1.4161 (lit.)

bp

202-203 °C (lit.)

mp

−4 °C (lit.)

solubility

H2O: slightly soluble
diethyl ether: easily soluble
ethanol: easily soluble

density

0.927 g/mL at 25 °C (lit.)

functional group

carboxylic acid

Organoleptic

pungent

SMILES string

CCCCCC(O)=O

InChI

1S/C6H12O2/c1-2-3-4-5-6(7)8/h2-5H2,1H3,(H,7,8)

InChI key

FUZZWVXGSFPDMH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexanoic acid is a resistance priming inducer that protects tomato plants from Botrytis cinerea[1].

Application

Hexanoic acid can be used:
  • To prepare polyethylene glycol derived nanomicelles as a non-viral gene carrier.[2]
  • As a starting material to synthesize an amino acid L-norleucine.[3]
  • As a reactant to prepare 4-methoxy phenyl hexyl ketone through Friedel−Crafts acylation of anisole using Hβ zeolite-based catalyst.[4]
  • As a template in the preparation of highly conducting thin polypyrrole coated alumina composite particles.[5]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective biocatalytic formal α-amination of hexanoic acid to l-norleucine
Dennig A, et al.
Organic & Biomolecular Chemistry, 16(43), 8030-8033 (2018)
Buddhadev Layek et al.
Molecular pharmaceutics, 11(3), 982-994 (2014-02-07)
Gene therapy holds immense potential as a future therapeutic strategy for the treatment of numerous genetic diseases which are incurable to date. Nevertheless, safe and efficient gene delivery remains the most challenging aspects of gene therapy. To overcome this difficulty
Friedel-Crafts acylation of anisole with hexanoic acid catalyzed by Hβ zeolite-supported tungstophosphoric acid
Bai G, et al.
Research on Chemical Intermediates, 41(8), 5041-5048 (2015)
Hexanoic acid and polyethylene glycol double grafted amphiphilic chitosan for enhanced gene delivery: influence of hydrophobic and hydrophilic substitution degree
Layek B, et al.
Molecular Pharmaceutics, 11(3), 982-994 (2014)
Formation of ultrathin polypyrrole (PPY) films on alumina particles using adsorbed hexanoic acid as a template.
Cho G, et al.
Langmuir, 16(10), 4424-4429 (2000)

Questions

1–4 of 4 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. What is the molarity of hexanoic acid?

    1 answer
    1. Based on density of 0.927 g/mL and MW of 116.16, the effective molarity of this neat liquid is 7.98 M.

      Helpful?

  4. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service