Skip to Content
Merck
All Photos(2)

Key Documents

P1386

Sigma-Aldrich

Propionic acid

≥99.5%

Synonym(s):

Acid C3, Propanoic acid, Propanyl acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2COOH
CAS Number:
Molecular Weight:
74.08
Beilstein:
506071
EC Number:
MDL number:
UNSPSC Code:
12352106
eCl@ss:
39021305
PubChem Substance ID:
NACRES:
NA.21

vapor density

2.55 (vs air)

Quality Level

vapor pressure

2.4 mmHg ( 20 °C)

Assay

≥99.5%

autoignition temp.

955 °F

expl. lim.

12.1 %

impurities

≤0.25% water (Karl Fischer)

color

≤10, APHA:

refractive index

n20/D 1.386 (lit.)

bp

141 °C (lit.)

mp

−24-−23 °C (lit.)

density

0.993 g/mL at 25 °C (lit.)

SMILES string

CCC(O)=O

InChI

1S/C3H6O2/c1-2-3(4)5/h2H2,1H3,(H,4,5)

InChI key

XBDQKXXYIPTUBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Propionic acid (PA) is a naturally occurring carboxylic acid, which in its pure state exists as a colorless corrosive liquid with an unpleasant odor. It is miscible in water. Industrially it is produced by hydrocarboxylation of ethylene in presence nickel carbonyl as a catalyst. PA has been found to reduce food intake, lower the fatty acids content in plasma and liver, might improve tissue insulin sensitivity and exerts immunosuppressive actions. Propionic acid is an excellent raw material as it is stable, cheap and safe and can even be used as a food additive.

Application


  • Distribution Characteristics of Low-Molecular-Weight Organic Acids in Reclaimed Soil Filled with Fly Ash: A Study.: This research investigates the presence and distribution of propionic acid among other organic acids in reclaimed soils treated with fly ash. The study provides insights into the environmental impact and potential remediation strategies for soils contaminated with industrial by-products (Zheng et al., 2024).

  • Effects of Black Soldier Fly (Hermetia illucens L., BSF) Larvae Addition on In Vitro Fermentation Parameters of Goat Diets.: This study examines how the addition of black soldier fly larvae affects the fermentation process of goat diets, including the production of propionic acid. The findings suggest potential benefits for improving animal nutrition and gut health (Lu et al., 2024).

  • NMR Spectroscopy in Diagnosis and Monitoring of Methylmalonic and Propionic Acidemias.: This review highlights the use of NMR spectroscopy to diagnose and monitor methylmalonic and propionic acidemias, providing a non-invasive method for tracking these metabolic disorders. The study underscores the clinical significance of propionic acid in metabolic research (Deleanu & Nicolescu, 2024).

  • The New Buffer Salt-Protected Sodium Butyrate Promotes Growth Performance by Improving Intestinal Histomorphology, Barrier Function, Antioxidative Capacity, and Microbiota Community of Broilers.: This study explores how propionic acid, along with sodium butyrate, improves the growth performance and gut health of broilers. The research demonstrates the beneficial effects of these acids on poultry farming (Melaku et al., 2024).

  • Prevotella enterotype associates with diets supporting acidic faecal pH and production of propionic acid by microbiota.: This study links the presence of the Prevotella enterotype in the gut microbiota to diets that promote the production of propionic acid, emphasizing the role of diet in modulating gut health and metabolic functions (Adamberg & Adamberg, 2024).


Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jialin Liu et al.
BMC biology, 18(1), 129-129 (2020-09-21)
The evolution of embryological development has long been characterized by deep conservation. In animal development, the phylotypic stage in mid-embryogenesis is more conserved than either early or late stages among species within the same phylum. Hypotheses to explain this hourglass
Kaori Watanabe et al.
Cell reports, 28(10), 2594-2607 (2019-09-05)
During evolution, organisms have acquired variable feeding habits. Some species are nutritional generalists that adapt to various food resources, while others are specialists, feeding on specific resources. However, much remains to be discovered about how generalists adapt to diversified diets.
Maria Litovchenko et al.
Science advances, 7(5) (2021-01-31)
Natural genetic variation affects circadian rhythms across the evolutionary tree, but the underlying molecular mechanisms are poorly understood. We investigated population-level, molecular circadian clock variation by generating >700 tissue-specific transcriptomes of Drosophila melanogaster (w1118 ) and 141 Drosophila Genetic Reference
Maria Elena Martino et al.
Cell host & microbe, 24(1), 109-119 (2018-07-17)
Animal-microbe facultative symbioses play a fundamental role in ecosystem and organismal health. Yet, due to the flexible nature of their association, the selection pressures that act on animals and their facultative symbionts remain elusive. Here we apply experimental evolution to
Flour and Breads and their Fortification in Health and Disease Prevention.
Victor R. Preedy, Ronald Ross Watson, Vinood B
Technology & Engineering, 476-476 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service