Merck
  • Europium bis(dimethylsilyl)amides including mixed-valent Eu3[N(SiHMe2)2]6[μ-N(SiHMe2)2]2.
Europium bis(dimethylsilyl)amides including mixed-valent Eu3[N(SiHMe2)2]6[μ-N(SiHMe2)2]2.
Dalton transactions (Cambridge, England : 2003) (2014-10-17)
André M Bienfait, Christoph Schädle, Cäcilia Maichle-Mössmer, Karl W Törnroos, Reiner Anwander
ABSTRACT
Trivalent Eu[N(SiHMe2)2]3(THF)2 can easily be synthesized by applying a routine salt metathesis protocol (EuCl3(THF)2 and 3 equiv. of Li[N(SiHMe2)2] in n-hexane) which crystallizes isotypically to its analogues of the rare-earth metal series (space group P21/c). Transsilylamination of Eu[N(SiMe3)2]2(THF)2 with a slight excess of HN(SiHMe2)2 in n-hexane-THF yields the divalent trinuclear compound Eu{[μ-N(SiHMe2)2]2Eu[N(SiHMe2)2](THF)}2, the solid-state structure of which differs significantly from the samarium and ytterbium analogues by showing three unique molecules in the asymmetric unit of which one is related to the two others by an inversion. Using crude Eu[N(SiMe3)2]3 in transsilylamination reactions with HN(SiHMe2)2 in n-hexane afforded n-hexane-insoluble trivalent ate complexes {MEu[N(SiHMe2)2]4}n (M = Na, K) depending on the synthesis conditions of Eu[N(SiMe3)2]3. Performing the transsilylamination of Eu[N(SiMe3)2]3 with a large excess of HN(SiHMe2)2 at elevated temperatures gave reproducibly the donor-free, mixed-valent, trinuclear compound Eu(II){[μ-N(SiHMe2)2]Eu(III)[N(SiHMe2)2]3}2 in good yield.
Sigma-Aldrich
Potassium bis(trimethylsilyl)amide, 95%
Sigma-Aldrich
Europium(III) chloride, powder, ≥99.9% trace metals basis
Sigma-Aldrich
n-Butyllithium solution, 2.5 M in hexanes
Sigma-Aldrich
n-Butyllithium solution, 10.7 M in hexanes
Sigma-Aldrich
n-Butyllithium solution, 2.0 M in cyclohexane
Sigma-Aldrich
n-Butyllithium solution, 1.6 M in hexanes
Sigma-Aldrich
Sodium amide, 95%
Sigma-Aldrich
Potassium bis(trimethylsilyl)amide solution, 1 M in THF
Sigma-Aldrich
Potassium bis(trimethylsilyl)amide solution, 0.5 M in toluene
Sigma-Aldrich
1,2-Diiodoethane, 99%
Sigma-Aldrich
Europium(III) chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium amide, 98%
Sigma-Aldrich
n-Butyllithium solution, 2.7 M in heptane
Sigma-Aldrich
Potassium bis(trimethylsilyl)amide solution, 1.0 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Potassium bis(trimethylsilyl)amide solution, 1.0 M in methyl tert-butyl ether
Europium, wire reel, 100mm, diameter 1.0mm, hard, 99.95%
Sigma-Aldrich
Diethyl ether
Supelco
Diethyl ether, analytical standard
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Diethyl ether, reagent grade, ≥98%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Diethyl ether, contains BHT as inhibitor, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Diethyl ether, puriss., contains ~5 mg/L 2,6-di-tert.-butyl-4-methylphenol as stabilizer, meets analytical specification of Ph. Eur., BP, ≥99.5% (GC)
Sigma-Aldrich
Diethyl ether, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Diethyl ether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, suitable for HPLC, ≥99.9%, inhibitor-free